Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2′-Deoxyguanosine 5′-triphosphate trisodium salt solution
Synonym(s):
dGTP-Na
3
Empirical Formula (Hill Notation):
C
10
H
13
N
5
Na
3
O
13
P
3
CAS Number:
93919-41-6
Molecular Weight:
573.13
EC Number:
300-026-5
Products
Brand
Sigma-Aldrich (2)
Color
Formula Weight
Physical Form
Purity
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
D7170
2′-Deoxyguanosine 5′-triphosphate trisodium salt solution,
10 mM in H
2
O
Expand
View Pricing
D5038
2′-Deoxyguanosine 5′-triphosphate trisodium salt solution,
100 mM (pH 7)
Expand
View Pricing