
2,6-Dichlorobenzyl bromide

Synonym(s):
α-Bromo-2,6-dichlorotoluene, 1-Bromomethyl-2,6-dichlorobenzene
Linear Formula:
Cl2C6H3CH2Br
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing