
25-Hydroxyvitamin D3 solution

Synonym(s):
25-Hydroxycholecalciferol solution
Empirical Formula (Hill Notation):
C27H44O2
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Technique
Showing 1-3 of 3