Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2,2′-Azobis(2-methylpropionitrile)
Synonym(s):
α,α′
-Azoisobutyronitrile, AIBN, Azobisisobutyronitrile, Free radical initiator
Linear Formula:
(CH
3
)
2
C(CN)N=NC(CH
3
)
2
CN
CAS Number:
78-67-1
Molecular Weight:
164.21
EC Number:
201-132-3
Products
Brand
Sigma-Aldrich (3)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
441090
2,2′-Azobis(2-methylpropionitrile),
98%
Expand
View Pricing
755745
2,2′-Azobis(2-methylpropionitrile),
recrystallized from methanol, 99%
Expand
View Pricing
01-7220
2,2′-Azobis(2-methylpropionitrile),
SAJ first grade, ≥98.0%
Expand
View Pricing