Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1-Methylimidazole
Synonym(s):
1-Methylimidazole
Empirical Formula (Hill Notation):
C
4
H
6
N
2
CAS Number:
616-47-7
Molecular Weight:
82.10
EC Number:
210-484-7
Products
Product Category
building blocks (2)
Brand
Sigma-Aldrich (3)
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
M50834
1-Methylimidazole,
ReagentPlus
®
, 99%
Expand
View Pricing
8.05852
1-Methylimidazole,
for synthesis
Expand
View Pricing
336092
1-Methylimidazole,
≥99%, purified by redistillation
Expand
View Pricing