Skip to Content
Merck
1,3-Bis(3-cyanopropyl)tetramethyldisiloxane 97%

1,3-Bis(3-cyanopropyl)tetramethyldisiloxane

1,3-Bis(3-cyanopropyl)tetramethyldisiloxane 97%
Linear Formula:
[NC(CH2)3Si(CH3)2]2O
CAS Number:
Molecular Weight:
268.50

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing