Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,2-Dimethoxyethane
Synonym(s):
mono-Glyme, Dimethylglycol, Ethylene glycol dimethyl ether, Monoglyme
Linear Formula:
CH
3
OCH
2
CH
2
OCH
3
CAS Number:
110-71-4
Molecular Weight:
90.12
EC Number:
203-794-9
Products
Product Category
solvents (5)
Brand
Sigma-Aldrich (5)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Anhydrous (2)
analytical standard (1)
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
E27408
1,2-Dimethoxyethane,
ReagentPlus
®
, ≥99%, inhibitor-free
Expand
View Pricing
307432
1,2-Dimethoxyethane,
suitable for HPLC, 99.9%, inhibitor-free
Expand
View Pricing
259527
1,2-Dimethoxyethane,
anhydrous, 99.5%, inhibitor-free
Expand
View Pricing
5.89574
1,2-Dimethoxyethane,
99.5%, anhydrous
Expand
View Pricing
72405
1,2-Dimethoxyethane,
analytical standard
Expand
View Pricing
09-1625
1,2-Dimethoxyethane,
SAJ first grade, ≥99.0%
Expand
View Pricing