Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,2:5,6-Di-
O
-isopropylidene-α-D-glucofuranose
Synonym(s):
D-Glucose diacetonide, Diacetone-D-glucose, 1,2,5,6-diisopropylidene-D-glucose
Empirical Formula (Hill Notation):
C
12
H
20
O
6
CAS Number:
582-52-5
Molecular Weight:
260.28
EC Number:
209-486-0
Products
Product Category
building blocks (2)
carbohydrates (2)
Brand
Sigma-Aldrich (2)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Optical Isomer
Special Grade
Purum (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
31460
1,2:5,6-Di-
O
-isopropylidene-α-D-glucofuranose,
purum, ≥98.0% (TLC)
Expand
View Pricing
D7600
1,2:5,6-Di-
O
-isopropylidene-α-D-glucofuranose,
98%
Expand
View Pricing