Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,1,1,3,3,3-Hexafluoro-2-propanol
Synonym(s):
HFP, Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoro-2-propanol
Linear Formula:
(CF
3
)
2
CHOH
CAS Number:
920-66-1
Molecular Weight:
168.04
EC Number:
213-059-4
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (2)
Supelco (2)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
105228
1,1,1,3,3,3-Hexafluoro-2-propanol,
≥99%
Expand
View Pricing
18127
1,1,1,3,3,3-Hexafluoro-2-propanol,
for LC-MS LiChropur
™
Expand
View Pricing
8.04515
1,1,1,3,3,3-Hexafluoro-2-propanol,
for synthesis
Expand
View Pricing
52517
1,1,1,3,3,3-Hexafluoro-2-propanol,
for GC derivatization, LiChropur
™
, ≥99.8%
Expand
View Pricing