Skip to Content
Merck
All Photos(1)

Key Documents

1559006

USP

Prednisone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1,4-Pregnadiene-17α,21-diol-3,11,20-trione, 1-Cortisone, 17α,21-Dihydroxy-1,4-pregnadiene-3,11,20-trione, Dehydrocortisone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O5
CAS Number:
Molecular Weight:
358.43
Beilstein:
2065301
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

prednisone

manufacturer/tradename

USP

mp

236-238 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1C=C[C@@]2(C)C(CC[C@]([C@@](CC[C@@]3(C(CO)=O)O)([H])[C@]3(C)C4)([H])[C@]2([H])C4=O)=C1

InChI

1S/C21H26O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-15,18,22,26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,18+,19-,20-,21-/m0/s1

InChI key

XOFYZVNMUHMLCC-ZPOLXVRWSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Prednisone USP reference standard for specified quality tests and assay use.

Also used to prepare standard, standard stock and internal standard solutions for the assay and performance tests according to the given below monographs of United States Pharmacopeia (USP):
  • Prednisone
  • Prednisone Tablets
  • Prednisone Oral Solution
  • Hydrocortisone Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prednisone Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 45(3), 3672-3672 (2020)
Prednisone
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(2), 3670-3670 (2020)
Prednisone Oral Solution
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 30(1), 3671-3671 (2020)
Hydrocortisone Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(6), 2234-2234 (2020)
Ronald de Wit
European journal of cancer (Oxford, England : 1990), 41(4), 502-507 (2005-03-02)
Until now, the use of systemic chemotherapy for advanced androgen-independent prostate cancer has had very little to offer to patients. However, in 2004, two large randomised trials investigating docetaxel vs. mitoxantrone have both demonstrated survival improvements, and, in one of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service