Skip to Content
Merck
All Photos(1)

Key Documents

48114

Supelco

Pesticide-HC Calibration Standards Kit

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

Agency

EPA 8080,8081

feature

standard type calibration

packaging

pkg of 1 kit

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

room temp

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Includes 1 ml of the following solutions, each at 1000 μg/ml in isooctane unless otherwise noted.

Kit Components Only

Product No.
Description

  • Aroclor 1016 solution (48097) 1 mL

  • Aroclor 1221 solution (48098) 1 mL

  • Aroclor 1232 solution (44805) 1 mL

  • Aroclor 1242 solution (44806) 1 mL

  • Aroclor 1248 solution (44807) 1 mL

  • Aroclor 1254 solution (44808) 1 mL

  • Aroclor 1260 solution(44809) 1 mL

  • Aroclor 1262 solution (44810) 1 mL

  • Chlordane (48065-U) 1 mL

  • TCL Pesticides Mix (48913) 1 mL

  • Toxaphene (48103) 1 mL

See All (11)

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service