Skip to Content
Merck
All Photos(1)

Key Documents

442551

Supelco

Dibromoacetic acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2CHCOOH
CAS Number:
Molecular Weight:
217.84
Beilstein:
1700457
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

128-130 °C/16 mmHg (lit.)

mp

32-38 °C (lit.)

density

2.382 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

OC(=O)C(Br)Br

InChI

1S/C2H2Br2O2/c3-1(4)2(5)6/h1H,(H,5,6)

InChI key

SIEILFNCEFEENQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibromoacetic acid belongs to the category of haloacetic acids (HAAs) which are a class of disinfection by-products produced in water.

Application

Dibromoacetic acid (Br2CHCOOH, CAS Number 631-64-1) may be used as an analytical reference standard for the determination of the analyte in aqueous samples by various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of haloacetic acids in aqueous environments by solid-phase extraction followed by ion-pair liquid chromatography-electrospray ionization mass spectrometric detection.
Loos R and Barcelo D
Journal of Chromatography A, 938(1-2), 45-55 (2001)
Applications of ion chromatography with electrospray mass spectrometric detection to the determination of environmental contaminants in water.
Roehl R, et al.
Journal of Chromatography A, 956(1-2), 245-254 (2002)
Determination of TOCl, TOBr and TOI in drinking water by pyrolysis and off-line ion chromatography.
Hua G and Reckhow DA
Analytical and Bioanalytical Chemistry, 384(2), 495-504 (2006)
Determination of haloacetic acids in water by acidic methanol esterification-GC-ECD method.
Nikolaou AD, et al.
Water Research, 36(4), 1089-1094 (2002)
Eric R V Dickenson et al.
Environmental science & technology, 42(9), 3226-3233 (2008-06-05)
While it is known that resorcinol- and phenol-type aromatic structures within natural organic matter (NOM) react during drinking water chlorination to form trihalomethanes (THMs), limited studies have examined aliphatic-type structures as THM and haloacetic acid (HAA) precursors. A suite of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service