Skip to Content
Merck
All Photos(1)

Key Documents

40003

Supelco

Acrylonitrile solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H3N
CAS Number:
Molecular Weight:
53.06
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

Agency

EPA 8031

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

InChI

1S/C3H3N/c1-2-3-4/h2H,1H2

InChI key

NLHHRLWOUZZQLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

National Toxicology Program technical report series, (573)(573), 1-155 (2012-07-28)
Styrene-acrylonitrile trimer (SAN Trimer) is a mixture of isomers formed by the condensation of two moles of acrylonitrile and one mole of styrene and has a molecular weight of 210. The mixture is composed of two structural forms: 4-cyano-1,2,3,4-tetrahydro-a-methyl-1-naphthaleneacetonitrile (THNA
Holly Meredith Kolenda-Roberts et al.
Toxicologic pathology, 41(1), 98-108 (2012-07-24)
Twenty-eight spontaneously occurring glial tumors (previously diagnosed as astrocytomas, oligodendrogliomas, and gliomas) and eleven granular cell tumors (GCTs) were selected for evaluation using a panel of immunohistochemistry (IHC) stains (Ricinus communis agglutinin type 1 [RCA-1], ionized calcium-binding adapter molecule 1
Danish J Malik et al.
PloS one, 7(8), e43354-e43354 (2012-08-24)
Novel carbon materials have been prepared by the carbonization of acrylonitrile (AN)/divinylbenzene (DVB) suspension porous copolymers having nominal crosslinking degrees in the range of 30-70% and obtained in the presence of various amounts of porogens. The carbons were obtained by
Pedro de la Torre et al.
Molecules (Basel, Switzerland), 17(10), 12072-12085 (2012-10-23)
(E)-2-(benzo[d]thiazol-2-yl)-3-heteroarylacrylonitriles are described as a new class of selective inhibitors of acetylcholinesterase (AChE). The most potent compound in the series exhibited good AChE inhibitory activity (IC₅₀ = 64 µM). Compound 7f was found to be more selective than galanthamine in
Hongwei Hou et al.
Analytical biochemistry, 430(1), 75-82 (2012-08-08)
The acrylonitrile metabolites 2-cyanoethylmercapturic acid (CEMA) and 2-hydroxyethylmercapturic acid (HEMA) have been determined in human urine using an automated column-switching procedure. A diluted sample was centrifuged just prior to being injected into a reusable precolumn packed with a restricted access

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service