Skip to Content
Merck
All Photos(15)

Key Documents

ZRB1844

Sigma-Aldrich

Anti-CD20 Antibody, clone 1E10 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E10, recombinant monoclonal

description

1E10 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 33.08 kDa
observed mol wt ~34 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD20(931)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1E10 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects CD20. It targets an epitope within 16 amino acids from the second extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the second extracellular domain from the C-terminal half of human CD20.

Application

Quality Control Testing

Evaluated by Western Blotting in Ramos cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CD20 in Ramos cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CD20 in Daudi cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CD20 in Ramos cells.

Affinity Binding Assay: A representative lot of this antibody bound CD20 with a KD of 3.2 x 10-7in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CD20 in human tonsil tissue sections.

Flow Cytometry Analysis: 1 μg from a representative lot detected CD20 in one million Ramos cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

B-lymphocyte antigen CD20 (UniProt: P11836; also known as B-lymphocyte surface antigen B1, Bp35, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, CD20) is encoded by the MS4A1 (also known as CD20) gene (Gene ID: 931) in human. CD20 is a multi-pass homotetrameric, non-glycosylated membrane protein that is expressed on the surface of normal and malignant B-lymphocytes. Its expression is reportedly initiated at the pre-B cell stage of development and remains present until terminal differentiation into a plasma cell at which time it is lost. It has three cytoplasmic domains, four transmembrane domains, and two extracellular domains. Its both N- and C-termini reside within the cytoplasm. It plays a role in the regulation of cellular calcium influx necessary for the development, differentiation, and activation of B-lymphocytes. It functions as a store-operated calcium channel component. It undergoes phosphorylation by PKC on serine and threonine residues in resting cells and these phosphorylations are reported to be higher in proliferating malignant B cells than in resting cells. CD20 is considered as an important therapeutic target in the treatment of B cell malignancies and various antibodies have been developed that bind to different epitopes. Mutations in MS4A1 gene are known to cause common variable immunodeficiency 5 (CVID5) that is characterized by failure of B-cell differentiation, hypogammaglobulinemia, recurrent bacterial infections, and an inability to mount an antibody response to antigen. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Pavlasova, G., and Mraz, M. (2020). Haematologica. 105(6); 1494-1506; Klein, C., et al. (2013). MAbs. 5(1); 22-33; Bubien, JK., et al. (1993). J. Cell Biol. 121(5); 1121-1132).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service