Z742654
Tube-O-DIALYZER™ mini dialysis system
Mixed, MWCO 8 K
Synonym(s):
semi-permeable membrane tube, single use tube dialysis
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
feature
Mixed
packaging
pkg of 20 ea
manufacturer/tradename
G-BioSciences 786-622
pore size
8 K MWCO
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Dialysis is a popular technique used for the exchange of buffer medium across semi-permeable membranes. Dialysis devices are available in many configurations for research applications. We offer innovative dialysis devices and accessories for processing small samples. The Tube-O-DIALYZER mini dialysis system is designed for small sample volumes of 20μl to 2.5ml.
The Tube-O-DIALYZER tube comes in two ideal sizes; the Micro unit allows efficient dialysis of 20-250μl samples and the Medi unit is optimized for samples between 0.2-2.5ml. Both sizes are available with membranes with molecular weight cutoffs (MWCO) of 1k, 4k, 8k, 15k, and 50k Daltons.
Tube-O-DIALYZER comes in packs of 20, each supplied with 6 appropriately sized floats and storage caps to allow storage of dialyzed samples. For added convenience, Tube-O-DIALYZER is also available as a mixed kit containing 10 Micro and 10 Medi units.
The Tube-O-DIALYZER tube comes in two ideal sizes; the Micro unit allows efficient dialysis of 20-250μl samples and the Medi unit is optimized for samples between 0.2-2.5ml. Both sizes are available with membranes with molecular weight cutoffs (MWCO) of 1k, 4k, 8k, 15k, and 50k Daltons.
Tube-O-DIALYZER comes in packs of 20, each supplied with 6 appropriately sized floats and storage caps to allow storage of dialyzed samples. For added convenience, Tube-O-DIALYZER is also available as a mixed kit containing 10 Micro and 10 Medi units.
Application
- Dialysis of small sample volumes
- Equilibrium dialysis for buffer exchange
- Dialysis for single use applications, such as infectious and radioactive samples
Legal Information
Tube-O-DIALYZER is a trademark of Geno Technology, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service