Skip to Content
MilliporeSigma

Skip To

WORKFLOW

IROA TruQuant Yeast Extract Workflow Kit

Supplied by IROA Technologies

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
$412.00

About This Item

NACRES:
NA.28
UNSPSC Code:
41116107

$412.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

IROA Technologies

application(s)

advanced drug delivery

shipped in

dry ice

storage temp.

−70°C

Application

TruQuant IQQ is a high-quality quantitation system for making simultaneous accurate biological measurements on several hundred biochemicals in small quantities of biological samples. This is achieved by (1) spiking a complex Internal Standard (WORKFLOW-A) into a biological sample to a) quantify all the biochemicals in the sample relative to their counterparts in the Internal Standard, b) suppression-correct each compound and c) normalize sample to sample variances; and (2) injecting the same well characterized Long-Term Reference Standard (WORKFLOW-B) to create a daily retention time (RT) library of all compounds to be found in the Internal Standard for reproducible ID, and to measure day-to-day (QA/QC) to assure reproducible instrument performance. The system is completely automated using IROA ClusterFinder software.

IROA TruQuant IQQ Workflow Kit contains the materials and tools for the analysis of 90 experimental samples. The kit is intended to be used for mass spectrometry metabolomics applications.

Features and Benefits

Identification: Confidence in identifying common and new metabolites
Quantification: Comprehensive correction of analytical and biological variability for all analytes
Quality Control: Convenient and automated tools for assuring reproducibility within/across studies

  • Simultaneous metabolite identification, quantitation and platform QC
  • Accurate compound formula ID for MS alone; complete ID with addition of SWATH, or IM
  • IROA patterns discriminate peaks of biological origin from artefactual peaks allowing the removal of false data
  • IROA-based fragments have the IROA ratio pattern of their parent peaks and can be identified as such using the “peak correlation” ClusterFinder module
  • Retention Index within the sample allows “unknowns” to be tracked with precision
  • Suppression-corrected measurements for significantly better quantitation
  • Normalizes (suppression-corrected) experimental samples to overcome day-to-day, instrument-to-instrument variances
  • ClusterFinder software solution builds libraries, IDs/quantitates compounds and normalizes data
  • Ensures high level QC for accurate and reproducible results

Other Notes

ClusterFinder software package is distributed with and is tailored to work with the IROA TruQuant IQQ kit. A user manual and video instructions are provided.
To download the software package, click here - ClusterFinder software.
Link to the ClusterFinder Videos below:


To access the MS Calculator Cascade Spreadsheet, follow the link below:
MS Calculator
For additional information on our range of Biochemicals, please complete this form.

Legal Information

ClusterFinder is a trademark of IROA Technologies LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ML0010OAMLSMSMLS
application(s)

advanced drug delivery

application(s)

advanced drug delivery

application(s)

-

application(s)

advanced drug delivery
lipidomics
metabolomics
microbiology

manufacturer/tradename

IROA Technologies

manufacturer/tradename

-

manufacturer/tradename

IROA Technologies

manufacturer/tradename

IROA Technologies

shipped in

dry ice

shipped in

-

shipped in

dry ice

shipped in

dry ice

storage temp.

−70°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters