Skip to Content
Merck
All Photos(1)

Key Documents

W1895

Sigma-Aldrich

WAY-100135

≥98% (HPLC)

Synonym(s):

3-[4-(2-Methoxyphenyl)piperazin-1-yl]-2-phenyl-N-tert-butyl-propanamide dihydrochloride;, N-tert-Butyl-3-(4-(2-methoxyphenyl)-piperazin-1-yl) -2-phenylpropanamide dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H33N3O2 · 2HCl
CAS Number:
Molecular Weight:
468.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

H2O: ≥10 mg/mL

originator

Wyeth

storage temp.

2-8°C

SMILES string

Cl.Cl.COc1ccccc1N2CCN(CC2)CC(C(=O)NC(C)(C)C)c3ccccc3

InChI

1S/C24H33N3O2.2ClH/c1-24(2,3)25-23(28)20(19-10-6-5-7-11-19)18-26-14-16-27(17-15-26)21-12-8-9-13-22(21)29-4;;/h5-13,20H,14-18H2,1-4H3,(H,25,28);2*1H

InChI key

VJGZNBYDSDEOED-UHFFFAOYSA-N

Biochem/physiol Actions

WAY-100135 is a potent, selective 5-HT1A receptor antagonist (IC50 = 15 nM). It is selective over 5-HT1B, 1C, 2,α1, α2 and D2 receptors (IC50 > 1000 nM).

Features and Benefits

This compound was developed by Wyeth. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kangjian Xiang et al.
Current neurovascular research, 13(4), 321-328 (2016-10-26)
Serotonin (5-hydroxytryptamine, 5-HT) is an important neurotransmitter that modulates N-methyl-D-aspartate (NMDA) receptor activity by binding to several different 5-HT receptor subtypes. In the present study, we used whole-cell patch-clamp recordings in transverse slice preparations to test the role of 5-HT
Daniel Sobrido-Cameán et al.
Disease models & mechanisms, 12(2) (2019-02-03)
Classical neurotransmitters are mainly known for their roles as neuromodulators, but they also play important roles in the control of developmental and regenerative processes. Here, we used the lamprey model of spinal cord injury to study the effect of serotonin
Lei Han et al.
The Journal of physiology, 599(1), 253-267 (2020-10-03)
Chemogenetic activation of medial vestibular nucleus-projecting 5-HT neurons resulted in deficits in vestibular-mediated tasks, including negative geotaxis, balance beam and rota-rod tests. The 5-HT1A receptor mediates the vestibular-related behavioural effects of 5-HT in the vestibular nucleus. 5-HT1A receptor activation attenuated

Articles

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service