Skip to Content
Merck
All Photos(1)

Key Documents

U0250

Sigma-Aldrich

Methyl undecanoate

≥98.0% (GC)

Synonym(s):

Undecanoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9CO2CH3
CAS Number:
Molecular Weight:
200.32
Beilstein:
1762129
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.429 (lit.)

density

0.872 g/mL at 25 °C (lit.)

functional group

ester

lipid type

saturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCC(=O)OC

InChI

1S/C12H24O2/c1-3-4-5-6-7-8-9-10-11-12(13)14-2/h3-11H2,1-2H3

InChI key

XPQPWPZFBULGKT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jana Orsavova et al.
International journal of molecular sciences, 16(6), 12871-12890 (2015-06-10)
Characterizations of fatty acids composition in % of total methylester of fatty acids (FAMEs) of fourteen vegetable oils--safflower, grape, silybum marianum, hemp, sunflower, wheat germ, pumpkin seed, sesame, rice bran, almond, rapeseed, peanut, olive, and coconut oil--were obtained by using

Protocols

Protocol for GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service