Skip to Content
Merck
All Photos(2)

Key Documents

T6778

Sigma-Aldrich

Anti-Rabbit IgG (whole molecule)–TRITC antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Rabbit IgG (whole molecule)–Tetramethylrhodamine isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

TRITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

contains

≥15 mM sodium azide as preservative

storage condition

protect from light

technique(s)

direct immunofluorescence: 1:400
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:400

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Binds all rabbit Igs.
IgG consists of four isotypes IgG1, IgG2, IgG3 and IgG4. It contains four polypeptide chains, made of two identical 50 kDa γ heavy (H) chains and two identical 25 kDa κ or λ light (L) chains, which is linked together by inter-chain disulfide bonds.

Application

Anti-Rabbit IgG (whole molecule)−TRITC antibody produced in goat has been used in immunocytochemistry.
Conjugation of TRITC to Rabbit IgG (whole molecule) antibody enables the use in immunofluorescent techniques.

Biochem/physiol Actions

IgG activates the classical pathway of the complement system. It neutralizes virus particles and toxins. IgG plays a key role in antibody-dependent cell-mediated cytotoxicity (ADCC) and phagocytosis. It might be associated with allergy. IgG mediates transplacental transport and participates in the secondary immune response. It has the longest serum half life.
IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.

Other Notes

Antibody adsorbed with human IgG.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4.

Preparation Note

Adsorbed to reduce background staining with human samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jesús Pintor et al.
Purinergic signalling, 1(1), 83-90 (2008-04-12)
Nucleotides present an important role in ocular physiology which has been demonstrated by recent works that indicate their involvement in many ocular processes. P2Y are important among P2 receptors since they can control tear production, corneal wound healing, aqueous humour
Kamin J Johnson et al.
Biology of reproduction, 66(4), 992-1000 (2002-03-22)
In the seminiferous epithelium, morphologically diverse junctions mediate inter-Sertoli and Sertoli-germ cell adhesive contact and likely transmit signals between contacting cells. Defining the molecular composition of testicular cell-cell junctions is an important step in determining their function. Proteins belonging to
Seunghyun Lee et al.
Scientific reports, 7(1), 14203-14203 (2017-10-29)
We investigated serial changes of the Kupffer cell (KC) function and hepatic oxygen saturation (sO
Tao Tao et al.
American journal of respiratory cell and molecular biology, 35(6), 668-680 (2006-07-01)
Antiinflammatory effects of glucocorticoids are critical to treatment of airway inflammation in such common disorders as asthma. There is considerable variation in responsiveness to glucocorticoid, and prolonged exposure can result in glucocorticoid resistance. We cloned LGL2, a glucocorticoid-inducible gene in
5-HT1B and 5-HT1D receptors in the human trigeminal ganglion: co-localization with calcitonin gene-related peptide, substance P and nitric oxide synthase
Hou M, et al.
Brain Research, 909(1-2), 112-120 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service