Skip to Content
Merck
All Photos(1)

Key Documents

T5898

Sigma-Aldrich

L-Threonine methyl ester hydrochloride

≥98% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH(NH2)COOCH3 · HCl
CAS Number:
Molecular Weight:
169.61
Beilstein:
3562717
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

L-Threonine methyl ester hydrochloride,

Assay

≥98% (TLC)

form

powder

color

white

mp

64 °C

storage temp.

−20°C

SMILES string

Cl.COC(=O)[C@@H](N)C(C)O

InChI

1S/C5H11NO3.ClH/c1-3(7)4(6)5(8)9-2;/h3-4,7H,6H2,1-2H3;1H/t3?,4-;/m0./s1

InChI key

OZSJLLVVZFTDEY-LXNQBTANSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geoffrey J Roff et al.
Journal of the American Chemical Society, 126(13), 4098-4099 (2004-04-01)
A series of diastereoisomers of beta-methyl-beta-phenylalanine analogues 1a-f have been prepared in enantiomerically pure form using a combination of chemo- and biocatalysis. Starting from l-threonine methyl ester 2, a range of beta,beta-disubstituted didehydroamino acids were obtained as their (Z)-isomers 6a-f.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service