Skip to Content
Merck
All Photos(1)

Key Documents

SRP3241

Sigma-Aldrich

IL-13 Long Form from rat

≥98% (SDS-PAGE & HPLC), recombinant, expressed in E. coli, lyophilized, suitable for cell culture

Synonym(s):

NC300 (Human) P600 (Murine), interleukin 13

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

product name

IL-13 Long Form from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

biological source

rat

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤5.0 ng/mL ED50

mol wt

12.3 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

IL13 (interleukin 13) is a classic cytokine produced by Th2 (T-helper) cells. It was originally identified as an inflammatory cytokine production-inhibiting T-cell derived cytokine.
Human and murine IL-13 is cross-species reactive. Recombinant rat IL-13 is a 11.9kDa protein consisting of 109 amino acid residues.

Biochem/physiol Actions

IL13 (interleukin 13) is up-regulated during parasitic infections, and participates in host′s protective immunity against Schistosoma japonicum. This protein promotes eosinophil activation resulting in elevated specific IgE against adult and larval antigens. During acute infection it promotes egg expulsion through increased intestinal contractility. It acts as a liver protective agent where it promotes formation of liver granulomas in a CD4+ T cell-mediated manner, which results in liver fibrosis. In airway disorders, such as asthma, chronic obstructive pulmonary disease (COPD), and cystic fibrosis lung disease, IL13 promotes the formation of airway goblet cells, resulting in excess mucus secretion, which in turn, is regulated by IL13-activated autophagy.

Sequence

TPGPVRRSTS PPVALRELIE ELSNITQDQK TSLCNSSIVW SVDLTAGGFC AALESLTNIS SCNAIHRTQR ILNGLCNQKA SDVASSPPDT KIEVAQFISK LLNYSKQLFR YGH

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IL-13 effector functions
Wynn TA
Annual Review of Immunology, 21, 425-456 (2003)
M Wills-Karp et al.
Science (New York, N.Y.), 282(5397), 2258-2261 (1998-12-18)
The worldwide incidence, morbidity, and mortality of allergic asthma are increasing. The pathophysiological features of allergic asthma are thought to result from the aberrant expansion of CD4(+) T cells producing the type 2 cytokines interleukin-4 (IL-4) and IL-5, although a
IL13 activates autophagy to regulate secretion in airway epithelial cells.
Dickinson JD et al
Autophagy, 12(2), 397-409 (2016)
An IL-13 promoter polymorphism associated with liver fibrosis in patients with Schistosoma japonicum.
Long X et al
PLoS ONE, 10(8), e0135360-e0135360 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service