Skip to Content
Merck
All Photos(1)

Key Documents

SML0581

Sigma-Aldrich

SR 58894A

≥98% (HPLC)

Synonym(s):

3-(2-Allylphenoxy)-1-[(1S)-1,2,3,4-tetrahydro-naphth-1-ylamino]-(2 S)-2-propanol hydrochloride, SR58894A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H27NO2 · HCl
CAS Number:
Molecular Weight:
373.92
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -19 to -25°, c = 1 in methanol

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

SR58894A is a potent and selective antagonist of β3-adrenergic receptor.
SR 58894A is also known as (3-(2-allylphenoxy)-1-[(1 S)-1,2,3,4-tetrahydronaphth-1-ylamino]-(2 S)-2-propanol hydrochloride, Kissei). It has the ability to antagonize relaxation, stimulated by isoprenaline.

Features and Benefits

This compound is featured on the β-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacological characterization of beta-adrenoceptor subtypes mediating relaxation in porcine isolated ureteral smooth muscle
Wanajo I et al.
The Journal of Urology, 172(3), 1155-1159 (2004)

Articles

β-Adrenoceptors are activated either by norepinephrine or epinephrine. Learn more about the role of β-adrenoceptors in protein expression, cell signaling, and metabolism research.

β-Adrenoceptors are activated either by norepinephrine or epinephrine. Learn more about the role of β-adrenoceptors in protein expression, cell signaling, and metabolism research.

β-Adrenoceptors are activated either by norepinephrine or epinephrine. Learn more about the role of β-adrenoceptors in protein expression, cell signaling, and metabolism research.

β-Adrenoceptors are activated either by norepinephrine or epinephrine. Learn more about the role of β-adrenoceptors in protein expression, cell signaling, and metabolism research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service