Skip to Content
Merck
All Photos(1)

Key Documents

SML0531

Sigma-Aldrich

BIO-Acetoxime

≥98% (HPLC)

Synonym(s):

(2′Z,3′E)-6-Bromoindirubin-3′-acetoxime, (3Z)-3-[(3E)-3-[(Acetyloxy)imino]-1,3-dihydro-2H-indol-2-ylidene]-6-bromo-1,3-dihydro-2H-Indol-2-one, 6-Bromoindirubin acetoxime, 6-Bromoindirubin-3′-acetoxime, BIA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12BrN3O3
CAS Number:
Molecular Weight:
398.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, dark red to purple

solubility

H2O: 2 mg/mL (clear solution, warmed)

storage temp.

2-8°C

SMILES string

CC(=O)O\N=C1\C(Nc2ccccc12)=C3\C(=O)Nc4cc(Br)ccc34

InChI

1S/C18H12BrN3O3/c1-9(23)25-22-16-12-4-2-3-5-13(12)20-17(16)15-11-7-6-10(19)8-14(11)21-18(15)24/h2-8,20H,1H3,(H,21,24)/b17-15-,22-16+

InChI key

MENDIXKFTXYTHJ-HPBPSMLHSA-N

Biochem/physiol Actions

BIO-Acetoxime is a potent and selective GSK-3a/b inhibitor that reduces invasiveness of gliomas and extends animal survival in intracranial glioma models. Also, BIO-Acetoxime induces Wnt signaling and inhibits CD8+ T cell effector differentiation.

Features and Benefits

This compound is featured on the GSK-3 page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jelle L G Haitsma Mulier et al.
PloS one, 13(6), e0197967-e0197967 (2018-06-12)
Acute kidney injury (AKI) complicates shock. Diagnosis is based on rising creatinine, a late phenomenon. Intrarenal vasoconstriction occurs earlier. Measuring flow resistance in the renal circulation, Renal Resistive Index (RRI), could become part of vital organ function assessment using Doppler
R S G Hutchings et al.
Journal of medical entomology, 53(6), 1330-1347 (2016-08-03)
The mosquito fauna (Culicidae) from remote northern areas of the State of Amazonas were sampled using Centers for Disease Control and Prevention, Shannon, Malaise, and Suspended traps, together with net sweeping and immature collections. One hundred and seven collections were
Ahmet Yalcin et al.
Geriatrics & gerontology international, 16(8), 903-910 (2015-08-08)
Sarcopenia is prevalent in older people, and is related to survival and disability. There are no data on sarcopenia evaluated according to European Working Group on Sarcopenia in Older People criteria for nursing home residents in Turkey. We aimed to
Role of Sex Hormones in the Control of Vegetative and Metabolic Functions of Middle-Aged Women.
Vincenzo Monda et al.
Frontiers in physiology, 8, 773-773 (2017-10-20)
Xinyan Bi et al.
Nutrients, 10(3) (2018-03-08)
Few studies have been conducted to measure body composition in Asian populations. In this study, we determined the percent body fat (PBF) by using dual-energy X-ray absorptiometry (DEXA), air-displacement plethysmography (ADP or BOD POD), bioelectrical impedance analysis (BIA) and skinfold

Articles

Glycogen synthase kinase 3 (GSK-3) is a highly conserved family of serine/threonine kinases for over 100 proteins in many pathways. See a list of accepted GSK-3 modulators and related products.

Glycogen synthase kinase 3 (GSK-3) is a highly conserved family of serine/threonine kinases for over 100 proteins in many pathways. See a list of accepted GSK-3 modulators and related products.

Glycogen synthase kinase 3 (GSK-3) is a highly conserved family of serine/threonine kinases for over 100 proteins in many pathways. See a list of accepted GSK-3 modulators and related products.

Glycogen synthase kinase 3 (GSK-3) is a highly conserved family of serine/threonine kinases for over 100 proteins in many pathways. See a list of accepted GSK-3 modulators and related products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service