Skip to Content
Merck
All Photos(2)

Key Documents

SML0245

Sigma-Aldrich

Candesartan cilexetil

≥98% (HPLC)

Synonym(s):

2-ethoxy-1-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1H-Benzimidazole-7-carboxylic acid 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, TCV 116, TCY 116

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H34N6O6
CAS Number:
Molecular Weight:
610.66
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥15 mg/mL

originator

Takeda

storage temp.

2-8°C

SMILES string

CCOc1nc2cccc(C(=O)OC(C)OC(=O)OC3CCCCC3)c2n1Cc4ccc(cc4)-c5ccccc5-c6nnn[nH]6

InChI

1S/C33H34N6O6/c1-3-42-32-34-28-15-9-14-27(31(40)43-21(2)44-33(41)45-24-10-5-4-6-11-24)29(28)39(32)20-22-16-18-23(19-17-22)25-12-7-8-13-26(25)30-35-37-38-36-30/h7-9,12-19,21,24H,3-6,10-11,20H2,1-2H3,(H,35,36,37,38)

InChI key

GHOSNRCGJFBJIB-UHFFFAOYSA-N

Gene Information

human ... AGTR1(185)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Candesartan cilexetil is the prodrug form of the potent angiotensin II receptor antagonist, candesartan. The prodrug is cleaved by esterases within the intestine to liberate the active molecule.

Features and Benefits

This compound is featured on the Angiotensin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Takeda. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2 Oral

Target Organs

Kidney,Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji-Young Jeon et al.
Drug development and industrial pharmacy, 39(9), 1296-1299 (2012-10-04)
Candesartan is a long-acting and selective nonpeptide AT1 subtype angiotensin II receptor antagonist. The aim of this study was to compare the pharmacokinetics and to evaluate the bioequivalence of two candesartan cilexetil 16 mg formulations. Forty healthy volunteers were randomly assigned
J M Mallion et al.
Journal of human hypertension, 14 Suppl 2, S33-S41 (2000-11-22)
Candesartan is one of the first angiotensin II receptor antagonists (AIIRAs) to be developed. It binds tightly to and dissociates slowly from the angiotensin subtype 1 (AT1) receptor in vitro. These binding characteristics differ from those of losartan, which demonstrates
Ezequiel Balmori Melian et al.
Drugs, 62(5), 787-816 (2002-04-04)
The combination of candesartan cilexetil [an angiotensin II type 1 (AT(1)) receptor antagonist] plus hydrochlorothiazide (a thiazide diuretic), has been used in the treatment of patients with hypertension. The blood pressure (BP) lowering effect of various doses of this combination
Stephanie E Easthope et al.
Drugs, 62(8), 1253-1287 (2002-05-16)
Candesartan cilexetil is converted to the angiotensin II receptor antagonist candesartan during absorption from the gastrointestinal tract. The selective and competitive binding of candesartan to the angiotensin II type 1 (AT(1)) receptor prevents binding of angiotensin II, a key mediator
Caroline Fenton et al.
Drugs, 65(4), 537-558 (2005-03-01)
Candesartan cilexetil is the orally administered pro-drug of candesartan, a highly selective antagonist of the angiotensin II subtype 1 receptor that mediates the pressor activities of angiotensin II. Candesartan cilexetil is widely used for the treatment of hypertension and has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service