Skip to Content
Merck
All Photos(1)

Key Documents

SMB00545

Sigma-Aldrich

Skullcapflavone II

≥90% (LC/MS-UV)

Synonym(s):

5-hydroxy-2-(2-hydroxy-6-methoxyphenyl)-6,7,8-trimethoxychromen-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18O8
CAS Number:
Molecular Weight:
374.34
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥90% (LC/MS-UV)

form

solid

solubility

DMSO: 1 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC1=C(C(C=C(C2=C(O)C=CC=C2OC)O3)=O)C3=C(OC)C(OC)=C1OC

InChI

1S/C19H18O8/c1-23-11-7-5-6-9(20)13(11)12-8-10(21)14-15(22)17(24-2)19(26-4)18(25-3)16(14)27-12/h5-8,20,22H,1-4H3

InChI key

GMQFOKBGMKVUQZ-UHFFFAOYSA-N

General description

Natrual product derived from plant source.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service