Skip to Content
Merck
All Photos(2)

Key Documents

SMB00339

Sigma-Aldrich

(-)-Tetrahydropalmatine

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25NO4
CAS Number:
Molecular Weight:
355.43
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

COC1=CC=C2C(CN(CCC3=C4C=C(OC)C(OC)=C3)[C@@]4([H])C2)=C1OC

InChI

1S/C21H25NO4/c1-23-18-6-5-13-9-17-15-11-20(25-3)19(24-2)10-14(15)7-8-22(17)12-16(13)21(18)26-4/h5-6,10-11,17H,7-9,12H2,1-4H3/t17-/m0/s1

InChI key

AEQDJSLRWYMAQI-KRWDZBQOSA-N

General description

Tetrahydropalmatine is an tetrahydroprotoberberine isoquinoline alkaloid found primarily in the Corydalis genus.

Biochem/physiol Actions

Tetrahydropalmatine is a potent muscle relaxant and has shown analgesic effects. Tetrahydropalmatine is known to help in the recovery of cocaine addiction, as it is an antagonist of dopamine (DA) D1 and D2 receptor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Levo-tetrahydropalmatine inhibits cocaine's rewarding effects: experiments with self-administration and brain-stimulation reward in rats
Xi ZX, et al.
Neuropharmacology, 53(6) (2007)
Levo-tetrahydropalmatine attenuates cocaine self-administration and cocaine-induced reinstatement in rats
Mantsch JR, et al.
Psychopharmacology, 192(4) (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service