Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702202

Sigma-Aldrich

Anti-MNX1/HB9/HLXB9 Antibody, clone 9P1B7, Rabbit Monoclonal

Synonym(s):

HB9, HLXB9, HOXHB9, SCRA1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

9P1B7, monoclonal

form

liquid

mol wt

41 kDa

species reactivity

mouse, mouse

concentration

0.30 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

SQQLLELEHQFKLNKYLSRPKRFEVATSLMLTETQVKIWFQNRRMKWKRSKKAKEQAAQEAEKQKGGGGGAGKGGAEEPGAEELLGPPAPGDKGSGRRLRD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MNX1(3110)

General description

This gene encodes a nuclear protein, which contains a homeobox domain and is a transcription factor. Mutations in this gene result in Currarino syndrome, an autosomic dominant congenital malformation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2009]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 250-350 of human MNX1/HB9/HLXB9 (P50219).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service