Skip to Content
Merck
All Photos(2)

Key Documents

SAB5200090

Sigma-Aldrich

Anti-Acetylated Lysine antibody produced in rabbit

1 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Acetylated Lysine, Anti-Lysine, Anti-acetyl lysine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

concentration

1 mg/mL

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: suitable

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

acetylation (Lys)

Specificity

Detects proteins containing acetylated lysine residues in SDS PAGE immunoblots.

Immunogen

Acetylated KLH Conjugated

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS, 50% glycerol, 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Egor A Vorontsov et al.
Molecular & cellular proteomics : MCP, 15(11), 3388-3404 (2016-11-03)
Protein post-translational methylation has been reported to occur in archaea, including members of the genus Sulfolobus, but has never been characterized on a proteome-wide scale. Among important Sulfolobus proteins carrying such modification are the chromatin proteins that have been described
Jingjing Cao et al.
Molecular microbiology (2018-04-22)
Nucleic acid-binding proteins of the Sac10b family, also referred to as Alba (for acetylation lowers binding affinity), are highly conserved in Archaea. It was reported that Sso10b, a Sac10b homologue from Sulfolobus solfataricus, was acetylated at the ɛ-amino group of
Michal Shilian et al.
Frontiers in cell and developmental biology, 10, 1015125-1015125 (2022-11-18)
The six subunits (Elp1 to Elp6) Elongator complex promotes specific uridine modifications in tRNA's wobble site. Moreover, this complex has been indirectly involved in the regulation of α-tubulin acetylation in microtubules (MTs) via the stabilization of ATP-Citrate Lyase (Acly), the
Yan Li et al.
International journal of molecular medicine, 41(6), 3517-3526 (2018-03-14)
Mitochondrial dynamics have critical roles in aging, and their impairment represents a prominent risk factor for myocardial dysfunction. Mitochondrial deacetylase sirtuin (SIRT)3 contributes greatly to the prevention of redox stress and cell aging. The present study explored the role of SIRT3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service