Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700455

Sigma-Aldrich

Monoclonal Anti-CD140a-APC antibody produced in mouse

clone 16A1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-PDGF-RA, Anti-PDGFRA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

Allophycocyanin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

16A1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PDGFRA(5156)

General description

Cluster of differentiation 140a (CD140a), also known as platelet-derived growth factor receptor α(PDGFRA), is encoded by the gene mapped to human chromosome 4q12. The encoded protein belongs to the receptor tyrosine kinase gene family.
The mouse monoclonal antibody 16A1 recognizes CD140a / PDGF-RA, the 170 kDa alpha chain of platelet-derived growth factor receptor, which is widely expressed on a variety of mesenchymal-derived cells and plays pro-proliferative or anti-proliferative roles in various tumours.

Immunogen

CD140a-transfected NIH 3T3 cells

Application

The reagent is designed for Flow Cytometry analysis of human blood cells using 10 μL reagent / 100 μL of whole blood or 1e6 cells in a suspension. The content of a vial (0.25 mL) is sufficient for 25 tests.

Biochem/physiol Actions

Platelet-derived growth factor receptor plays a vital role in the development and maturation of platelets. Cluster of differentiation 140a (CD140a)/ PDGFRA acts as a potential target of for imatinib, a revolutionary drug for the treatment of chronic myeloid leukemia. PDGFRA pathway might be involved in developmental process of thrombocytes. The encoded protein facilitates hepatic stellate cell (HSCs) proliferation and migration. Mutation in the gene has been observed in gastrointestinal stromal tumors (GISTs).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 15 mM sodium azide and 0.2% high-grade protease free BSA as a stabilizing agent.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Platelet-Derived Growth Factor Receptor α Contributes to Human Hepatic Stellate Cell Proliferation and Migration.
Kikuchi A, et al.
The American Journal of Pathology, 187 (2017)
A 1.8-Mb YAC contig spanning three members of the receptor tyrosine kinase gene family (Pdgfra, Kit, and Flk1) on mouse chromosome 5
Brunkow ME, et al.
Genomics, 25 (1995)
PDGFRα promoter polymorphisms and expression patterns influence risk of development of imatinib-induced thrombocytopenia in chronic myeloid leukemia: A study from India
Guru SA, et al.
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 39 (2017)
PDGFRA mutations in gastrointestinal stromal tumors: frequency, spectrum and in vitro sensitivity to imatinib.
Corless CL, et al.
Journal of Clinical Oncology, 23, 5357-5364 (2005)
Overexpressed Skp2 within 5p amplification detected by array-based comparative genomic hybridization is associated with poor prognosis of glioblastomas.
Saigusa K, et al.
Cancer Science, 96, 676-676 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service