Skip to Content
Merck
All Photos(3)

Key Documents

SAB4500120

Sigma-Aldrich

Anti-C/EBP-ε antibody produced in rabbit

affinity isolated antibody

Synonym(s):

C/EBP ε, CCAAT/enhancer binding protein ε, CEBPE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 30 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CEBPE(1053)

General description

Anti-C/EBP-ε Antibody detects endogenous levels of total C/EBP-ε protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human C/EBP-epsilon.

Immunogen Range: 40-89

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Sun et al.
Journal of cellular biochemistry, 119(3), 2891-2899 (2017-11-08)
Osteoarthritis (OA) is the leading degenerative joint disease and featured by articular cartilage destruction, where chondrocyte apoptosis plays a critical role. Semaphorin-3A (Sema3A) has been implicated in OA chondrocyte physiology. In this study we aimed to uncover how Sema3A signaling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service