Skip to Content
Merck
All Photos(1)

Key Documents

S7771

Sigma-Aldrich

(S)-(−)-Sulpiride

≥98% (titration)

Synonym(s):

(S)-5-Aminosulfonyl-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2-methoxybenzamide, Levosulpiride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23N3O4S
CAS Number:
Molecular Weight:
341.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (titration)

mp

183-186 °C (lit.)

originator

Sanofi Aventis

storage temp.

2-8°C

SMILES string

CCN1CCC[C@H]1CNC(=O)c2cc(ccc2OC)S(N)(=O)=O

InChI

1S/C15H23N3O4S/c1-3-18-8-4-5-11(18)10-17-15(19)13-9-12(23(16,20)21)6-7-14(13)22-2/h6-7,9,11H,3-5,8,10H2,1-2H3,(H,17,19)(H2,16,20,21)/t11-/m0/s1

InChI key

BGRJTUBHPOOWDU-NSHDSACASA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(−)-Sulpiride was used to study the effect of dopamine D2 receptor function on activation of KCNQ potassium channels10 and on depression-like behavior in ovariectomized female rats.11

Biochem/physiol Actions

(S)-(−)-Sulpiride or Levosulpiride blocks the inhibitory enteric D2 receptors (neuronal and muscular). It has prokinetic activity and is effective in the treatment of functional dyspepsia and gastroparesis in insulin-dependent diabetes mellitus patients.7,8 Since dopamine controls human sexual function, (S)-(−)-Sulpiride is reportedly effective in treatment of erectile dysfunctions.9
D2 dopamine receptor antagonist; antipsychotic.

Features and Benefits

This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuya Hikima et al.
Cell reports, 35(1), 108951-108951 (2021-04-08)
Somatodendritic dopamine (DA) release from midbrain DA neurons activates D2 autoreceptors on these cells to regulate their activity. However, the source of autoregulatory DA remains controversial. Here, we test the hypothesis that D2 autoreceptors on a given DA neuron in
Trine Ljungstrom et al.
Pflugers Archiv : European journal of physiology, 446(6), 684-694 (2003-06-27)
Activation of KCNQ potassium channels by stimulation of co-expressed dopamine D(2) receptors was studied electrophysiologically in Xenopus laevis oocytes and in mammalian cells. To address the specificity of the interaction between D(2)-like receptors and KCNQ channels, combinations of KCNQ1-5 channels
W Hauber et al.
Behavioural brain research, 106(1-2), 143-150 (1999-12-14)
In the present study, the involvement of dopamine D1 and D2 receptors in the dorsal globus pallidus (GP) in motor control was investigated in rats. Results show that bilateral microinfusions of the dopamine D1 receptor antagonist SCH23390 or the dopamine
In Bok Paek et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 809(2), 345-350 (2004-08-19)
A rapid, sensitive and selective hydrophilic interaction liquid chromatography-tandem mass spectrometric (HILIC-MS/MS) method for the determination of levosulpiride in human plasma was developed. Levosulpiride and internal standard, tiapride were extracted from human plasma with ethyl acetate at pH 11 and
Fermín Mearin et al.
Clinical gastroenterology and hepatology : the official clinical practice journal of the American Gastroenterological Association, 2(4), 301-308 (2004-04-07)
Levosulpiride is a benzamide derivate D(2) dopamine antagonist with prokinetic activity that can accelerate gastric emptying and reduce discomfort in response to gastric distention. The aim of the study is to compare the clinical efficacy of levosulpiride and cisapride in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service