S7764
Complement sera from rabbit
lyophilized powder
Synonym(s):
Rabbit Serum
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
rabbit
Quality Level
form
lyophilized powder
composition
protein, ≥40 mg/mL biuret
technique(s)
cell culture | embryo: suitable
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−20°C
Application
Complement sera from rabbit has been used:
- for the incubation of embryo prior to inner cell mass isolation
- in the complement-dependent cytotoxicity (CDC) assay of peripheral blood mononuclear cells (PBMCs)
- in the purification of schwann cells
Biochem/physiol Actions
Rabbit complement sera was shown to promote acrosomal loss in rabbit spermatozoa due to activation of the complement.
Physical form
Lyophilized powder from indicated amount of serum
Reconstitution
Reconstitute with ice cold deionized water using volume indicated by package size
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Evidence that LDL receptor-related protein 1 (LRP1) acts as an early injury detection receptor and activates c-Jun in Schwann cells
Neuroreport, 27(18), 1305-1305 (2016)
Conventional pluripotency markers are unspecific for bovine embryonic-derived cell-lines
Theriogenology, 69(9), 1159-1164 (2008)
PLoS pathogens, 16(12), e1009169-e1009169 (2020-12-29)
Human cytomegalovirus (HCMV) is the primary viral cause of congenital birth defects and causes significant morbidity and mortality in immune-suppressed transplant recipients. Despite considerable efforts in vaccine development, HCMV infection still represents an unmet clinical need. In recent phase II
Comparison of Physical Crossmatch and Virtual Crossmatch to Identify Preexisting Donor-Specific Human Leukocyte Antigen (HLA) Antibodies and Outcome Following Kidney Transplantation
Medical Science Monitor : International Medical Journal of Experimental and Clinical Research, 25(9), 952-952 (2019)
PloS one, 8(1), e49638-e49638 (2013-01-26)
Bacterial pathogens need to acquire nutrients from the host, but for many nutrients their importance during infection remain poorly understood. We have investigated the importance of methionine acquisition and synthesis for Streptococcus pneumoniae growth and virulence using strains with gene
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service