Skip to Content
Merck
All Photos(1)

Key Documents

S6047

Sigma-Aldrich

Saccharin sodium salt hydrate

BioXtra, ≥99%

Synonym(s):

2,3-Dihydro-3-oxobenzisosulfonazole sodium salt, 2-Sulfobenzoic acid imide sodium salt, o-Sulfobenzimide sodium salt, Saccharin soluble

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4NNaO3S · xH2O
CAS Number:
Molecular Weight:
205.17 (anhydrous basis)
Beilstein:
3599229
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Assay

≥99%

form

powder

sweetness

300 × sucrose

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.001%
NH4+: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

[Na+].[H]O[H].[O-]C1=NS(=O)(=O)c2ccccc12

InChI

1S/C7H5NO3S.Na.H2O/c9-7-5-3-1-2-4-6(5)12(10,11)8-7;;/h1-4H,(H,8,9);;1H2/q;+1;/p-1

InChI key

ILJOYZVVZZFIKA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Saccharin is an artificial sweetener, which is used in canned fruit juices, vegetables, cookies and jellies. It is associated with bladder cancer.

Application

Saccharin sodium salt hydrate has been used to eliminate the potentially confounding effect of caloric content. It has also been used to study its effects on the cyclamate-responsive taste 2 receptor member 1 (TAS2R1) receptor.

Biochem/physiol Actions

′First generation′ synthetic sugar alternative that has been studied for its carcinogenic potential.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

FGF21 regulates sweet and alcohol preference
Talukdar S, et al.
Cell Metabolism, 23(2), 344-349 (2016)
Blends of non-caloric sweeteners saccharin and cyclamate show reduced off-taste due to TAS2R bitter receptor inhibition
Behrens M, et al.
Cell Chemical Biology, 24(10), 1199-1204 (2017)
Robert A Boakes et al.
Physiology & behavior, 218, 112822-112822 (2020-02-01)
Rats first given 24-h access to 10% sucrose for 4 or 12 days (Stage 1) were then switched to a saccharin solution for a 12-day Stage 2. The initial result of this switch was that these Sucrose groups drank less
Simone Rehn et al.
Physiology & behavior, 207, 15-27 (2019-05-06)
When animals are given access to a palatable food or drink on some days but not on others, the amount they consume can far exceed the daily amounts consumed by controls given daily access. A previous study demonstrated such bingeing
Spectrophotometric method for the determination of saccharin in food and pharmaceutical products
Mathew SB, et al.
Indian Journal of Pharmaceutical Sciences, 68(6), 821-821 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service