Skip to Content
Merck
All Photos(4)

Key Documents

S5136

Sigma-Aldrich

Sodium phosphate dibasic

BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%

Synonym(s):

sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
38070211
PubChem Substance ID:
NACRES:
NA.25

product line

BioReagent

Assay

≥99.0%

form

powder or crystals

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

pH

8.9-9.2 (25 °C, 50 g/L in water)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

solubility

water: soluble 100 mg/mL

application(s)

diagnostic assay manufacturing

storage temp.

room temp

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sodium phosphate dibasic has been used:
  • in the preparation of 1X PBS (phosphate buffer saline) for whole organ decellularization of porcine kidneys
  • as a component of PBS in porcine pancreas decellularization
  • in the preparation of synthetic P calibration material
  • in the preparation of PBS (phosphate-buffered saline)
  • in the preparation of M9 buffer

related product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolating lipid droplets from multiple species
Ding Y, et al.
Nature Protocols, 8(1), 43-43 (2013)
Rapid and accurate analyses of silicon and phosphorus in plants using a portable X-ray fluorescence spectrometer
Reidinger S, et al.
The New phytologist, 195(3), 699-706 (2012)
Evaluation of Burkholderia cepacia complex bacteria pathogenicity using Caenorhabditis elegans
Tedesco P, et al.
Bio-protocol, 6(20) (2016)
Decellularization methods of porcine kidneys for whole organ engineering using a high-throughput system
Sullivan D C, et al.
Biomaterials, 33(31), 7756-7764 (2012)
Yan Li et al.
Structure (London, England : 1993), 26(4), 555-564 (2018-03-13)
Zika virus (ZIKV) infection has become a global public health concern. The viral NS2B-NS3 protease is an attractive antiviral target because of its role in maturation of viral non-structural proteins. Substrate-derived protease inhibitors have been investigated, but it remains challenging

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service