Skip to Content
Merck
All Photos(2)

Key Documents

RES3077T-A7

SAFC

Tricine

Pharma Manufacturing

Synonym(s):

Tricine, N-[Tris(hydroxymethyl)methyl]glycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2)3CNHCH2CO2H
CAS Number:
Molecular Weight:
179.17
Beilstein:
1937804
EC Number:
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.21

biological source

synthetic

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, microbial, and trace metals; tested

useful pH range

7.4-8.8

pKa (25 °C)

8.1

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

SMILES string

OCC(CO)(CO)NCC(O)=O

InChI

1S/C6H13NO5/c8-2-6(3-9,4-10)7-1-5(11)12/h7-10H,1-4H2,(H,11,12)

InChI key

SEQKRHFRPICQDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Buffer quality is vital for the success of biopharmaceutical processes, because buffers are indispensable in nearly every production step.

Our broad portfolio of buffer materials manufactured under appropriate controls is tailored to your needs. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Application

Buffer component for separation of low molecular weight peptides.
Tricine is a biological buffer often referred to as a “Good′s” buffer. The pKa of Tricine is 8.1 which makes Tricine is a good candidate for biological applications that require pH control above physiological. Tricine is considered to be non-toxic to culture cell lines, highly water soluble and provides high-solution clarity.

Tricine can be used in a wide array of biological applications including as a component in media formulations. Specific applications include buffers for electrophoresis, protein purification and diagnostic reagent production.

Packaging

Product is available in the following package sizes:
RES3077T-A701X: 100 gm container
RES3077T-A702X: 1 kg container
RES3077T-A704X: 10 kg container
RES3077T-A705X: 25 kg container

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Nilsson et al.
Electrophoresis, 31(3), 459-464 (2010-02-02)
Totally porous lipid-based liquid crystalline nanoparticles were used as pseudostationary phase for capillary electroseparation with LIF detection of proteins at physiological conditions using unmodified cyclic olefin copolymer capillaries (Topas, 6.7 cm effective length). In the absence of nanoparticles, i.e. in
Arpita Gantayet et al.
Biofouling, 29(1), 77-85 (2012-12-06)
The freshwater zebra mussel (Dreissena polymorpha) is a notorious biofouling organism. It adheres to a variety of substrata underwater by means of a proteinaceous structure called the byssus, which consists of a number of threads with adhesive plaques at the
Hermann Schägger
Nature protocols, 1(1), 16-22 (2007-04-05)
Tricine-SDS-PAGE is commonly used to separate proteins in the mass range 1-100 kDa. It is the preferred electrophoretic system for the resolution of proteins smaller than 30 kDa. The concentrations of acrylamide used in the gels are lower than in
Bin Deng et al.
PloS one, 9(7), e102362-e102362 (2014-07-19)
Maresins are a new family of anti-inflammatory and pro-resolving lipid mediators biosynthesized from docosahexaenoic acid (DHA) by macrophages. Here we identified a novel pro-resolving product, 13R,14S-dihydroxy-docosahexaenoic acid (13R,14S-diHDHA), produced by human macrophages. PCR mapping of 12-lipoxygenase (12-LOX) mRNA sequence in
Thierry Rabilloud
Journal of proteomics, 73(8), 1562-1572 (2010-04-17)
Electrophoretic separations of proteins are widely used in proteomic analyses, and rely heavily on SDS electrophoresis. This mode of separation is almost exclusively used when a single dimension separation is performed, and generally represents the second dimension of two-dimensional separations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service