Skip to Content
Merck
All Photos(1)

Key Documents

R8382

Sigma-Aldrich

Reagent Alcohol

used for histology tissue preparation

Synonym(s):

SD3A Alcohol, Specially Denatured Ethanol, 3A formulation, Ethanol, RGA, denatured alcohol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:
NACRES:
NA.47

biological source

synthetic

vapor density

1.59 (vs air)

form

solution

expl. lim.

3.1-27.7 % (lit.)

concentration

100%

color

colorless

bp

78 °C (lit.)

mp

-114 °C (lit.)

density

0.790 g/cm3

application(s)

hematology
histology

storage temp.

room temp

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Application

Reagent alcohol has been used in most histology and cytology procedures for histology tissue preparation.

Components

95 parts of specially denatured ethyl alcohol 3A with 5 parts of isopropyl alcohol. Final concentrations are ~ 90% ethanol, 5% methanol and 5% isopropanol.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DNA stable-isotope probing.
Neufeld J D, et al.
Nature Protocols, 2(4), 860-860 (2007)
Timothy R M, et al.
Seasonal Stock Contributions of the Inriver Run and Sport Harvest for Tributary and Mainstem Spawning Chinook Salmon in the Kenai River, Alaska. (2013)
U Allenstein et al.
Soft matter, 11(25), 5053-5059 (2015-06-02)
Cells require adhesion to survive, proliferate and migrate, as well as for wound healing and many other functions. The strength of contractile cell forces on an underlying surface is a highly relevant quantity to measure the affinity of cells to
Sonia Soloneski et al.
Toxicology in vitro : an international journal published in association with BIBRA, 29(5), 834-844 (2015-03-31)
In vitro effects of the carbamates pirimicarb and zineb and their formulations Aficida® (50% pirimicarb) and Azzurro® (70% zineb), respectively, were evaluated in Chinese hamster ovary (CHO-K1) cells. Whereas the cytokinesis-blocked micronucleus cytome assay was employed to test for genotoxicity
Nicholas J Niemuth et al.
Chemosphere, 135, 38-45 (2015-04-22)
The occurrence of intersex fish, where male reproductive tissues show evidence of feminization, have been found in freshwater systems around the world, indicating the potential for significant endocrine disruption across species in the ecosystem. Estrogens from birth control medications in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service