Skip to Content
Merck
All Photos(1)

Key Documents

PZ0015

Sigma-Aldrich

Trovafloxacin mesylate

>98% (HPLC), powder, Pannexin 1 inhibitor

Synonym(s):

(1α,5α,6α)-7-(6-Amino-3-azabicyclo[3.1.0]hex-3-yl)-1-(2,4-difluorophenyl)-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic acid methanesulfonate, CP-99219-27, Trovafloxacin methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H15F3N4O3 · CH3SO3H
CAS Number:
Molecular Weight:
512.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Trovafloxacin mesylate, >98% (HPLC)

Assay

>98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >10 mg/mL

storage temp.

room temp

SMILES string

CS(O)(=O)=O.NC1C2CN(CC12)c3nc4N(C=C(C(O)=O)C(=O)c4cc3F)c5ccc(F)cc5F

InChI

1S/C20H15F3N4O3.CH4O3S/c21-8-1-2-15(13(22)3-8)27-7-12(20(29)30)17(28)9-4-14(23)19(25-18(9)27)26-5-10-11(6-26)16(10)24;1-5(2,3)4/h1-4,7,10-11,16H,5-6,24H2,(H,29,30);1H3,(H,2,3,4)/t10-,11+,16+;

InChI key

DYNZICQDCVYXFW-AHZSKCOESA-N

Application

Trovafloxacin mesylate has been used as a test compound in toxicity assay to assess its toxicity in 2D hepatocyte cultures. It has also been used to manipulate the mechanism of apoptotic cell disassembly during apoptosis.

Biochem/physiol Actions

Trovafloxacin mesylate acts as a pannexin1 (Panx1) inhibitor.
Trovafloxacin mesylate is a broad spectrum antibiotic. Trovafloxacin mesylate blocks the activity of DNA gyrase and topoisomerase IV, enzymes essential in the repliction, transcription, and repair of bacterial DNA.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioprinted 3D primary liver tissues allow assessment of organ-level response to clinical drug induced toxicity in vitro
Nguyen DG, et al.
PLoS ONE, 11(7), e0158674-e0158674 (2016)
Determining the contents and cell origins of apoptotic bodies by flow cytometry
Jiang L, et al.
Scientific reports, 7(1), 14444-14444 (2017)
Akira Nakajima et al.
Cell biology and toxicology, 34(1), 65-77 (2017-03-13)
Fluoroquinolones and propionic acid derivatives are widely used antibacterials and non-steroidal anti-inflammatory drugs, respectively, which have been reported to frequently trigger drug hypersensitivity reactions. Such reactions are induced by inflammatory mediators such as cytokines and chemokines. The present study investigated
Jieyu Xu et al.
Toxicology in vitro : an international journal published in association with BIBRA, 48, 286-301 (2018-02-07)
Immortalized liver cells have been used for evaluating the toxicity of compounds; however, excessive glutathione is considered to lessen cytotoxicity. In this study, we compared the effects of glutathione depletion on cytotoxicities of drugs using HepaRG and HepG2 cells, which

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service