Skip to Content
Merck
All Photos(2)

Key Documents

P3902

Sigma-Aldrich

Anti-Pyk2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CAK-β, Anti-Proline rich Kinase 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 110-116 kDa

species reactivity

rat, mouse, human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100 using tissue sections of human cerebellum.
immunoprecipitation (IP): 3-5 μg/mL using 150-200 μg of PC-12 rat pheochromocytoma RIPA lysate
microarray: suitable
western blot: 1:2,000 using a whole extract of LPS-stimulated P388 mouse monocyte-macrophage cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTK2B(2185)
mouse ... Ptk2b(19229)
rat ... Ptk2b(50646)

General description

Pyk2 (proline-rich kinase 2) protein belongs to tyrosine protein kinase family and is primarily expressed in the central nervous system and in cells derived from hematopoietic lineages. Pyk2 is found in tissues and cells like mesenchymal, epithelial, endothelial cells, neonatal cardiomyocytes, osteoclasts and neuronal cells.

Immunogen

Synthetic peptide corresponding to amino acid residues 991-1009 of human Pyk2, coupled to KLH. This sequence is highly conserved in rat and mouse (1 amino acid substitution).

Application

Anti-Pyk2 antibody produced in rabbit has been used in:
  • immunocytochemistry
  • immunohistochemistry
  • western blotting

Biochem/physiol Actions

Protein tyrosine kinases (PTKs) are critical components of the signalling pathways that control cell growth, differentiation, apoptosis, metabolism, cell cycle regulation and cytoskeletal function. Pyk2 has been detected in cell-cell contacts, at focal adhesion-like structures and podosomes, cytoplasmic perinuclear region, in association with actin filaments and diffusely distributed in the cytoplasm. Pyk2 phosphorylation is critical for its interaction with SH2-containing signalling molecules and their linkage to signalling pathways that regulate extracellular-signal-regulated kinase (ERK), Janus kinases (JNK) and p38 kinases. Pyk2 has been shown to interact with Src family kinases, the growth factor receptor-bound protein 2 (Grb2)/Sos complex, p130cas, paxillin, Hic-5, and several other proteins, including inhibitors, to regulate signalling as well as cytoskeletal and morphological changes of cells. It has a crucial role in T and B cell antigen receptor signaling, cell cycle progression, metastasis, NK cytotoxicity, modulation of ion channel function and neuronal short- and long- term responses.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RAFTK/Pyk2-mediated cellular signalling
Avraham H, et al.
Cellular Signalling, 12(3), 123-133 (2000)
Shakir Hasan et al.
Toxins, 11(6) (2019-06-23)
Myeloid phagocytes have evolved to rapidly recognize invading pathogens and clear them through opsonophagocytic killing. The adenylate cyclase toxin (CyaA) of Bordetella pertussis and the edema toxin (ET) of Bacillus anthracis are both calmodulin-activated toxins with adenylyl cyclase activity that
Anirban Bhattachariya et al.
Physiological reports, 2(7) (2014-10-28)
Stretch of vascular smooth muscle stimulates growth and proliferation as well as contraction and expression of contractile/cytoskeletal proteins, all of which are also regulated by calcium-dependent signals. We studied the role of the calcium- and integrin-activated proline-rich tyrosine kinase 2
Pyk2 is essential for astrocytes mobility following brain lesion
Giralt A, et al.
Glia, 64(4), 620-634 (2016)
Vincenzo Mastrolia et al.
Scientific reports, 11(1), 16357-16357 (2021-08-13)
Pyk2 is a Ca2+-activated non-receptor tyrosine kinase enriched in the forebrain, especially in pyramidal neurons of the hippocampus. Previous reports suggested its role in hippocampal synaptic plasticity and spatial memory but with contradictory findings possibly due to experimental conditions. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service