Skip to Content
Merck
All Photos(2)

Key Documents

P1223

Sigma-Aldrich

L-α-Phosphatidylethanolamine, dioleoyl

≥99% (GC), ≥98% (TLC), lyophilized powder

Synonym(s):

1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, (9Z)-9-Octadecenoic acid (1R)-1-[[[(2-aminoethoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester, 1,2-Di(9Z-octadecenoyl)-sn-glycero-3-phosphoethanolamine, 3-sn-Phosphatidylethanolamine, 1,2-dideoyl, L-β,γ-Dioleoyl-α-cephalin, DOPE, Dioleoyl phosphatidylethanolamine, PE(18:1(9Z)/18:1(9Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H78NO8P
CAS Number:
Molecular Weight:
744.03
Beilstein:
1718431
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Quality Level

Assay

≥98% (TLC)
≥99% (GC)

form

lyophilized powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)OCC(COP(O)(=O)OCCN)OC(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C41H78NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-40(43)47-37-39(38-49-51(45,46)48-36-35-42)50-41(44)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20,39H,3-16,21-38,42H2,1-2H3,(H,45,46)/b19-17-,20-18-

InChI key

MWRBNPKJOOWZPW-CLFAGFIQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine (DOPE) forms heterogenous liposomes with N-[1-(2,3-dioleoyloxy)propyl]-N,N,N-trimethylammonium methylsulfate (DOTAP), which are used as delivery vehicles for therapeutic agents .
Forms liposomes with DC-cholesterol that can be used to efficiently transfect several mammalian cell lines.

Packaging

Packaged under Argon.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiao Wang et al.
Scientific reports, 8(1), 10795-10795 (2018-07-19)
P4-ATPases are a subfamily of P-type ATPases that flip phospholipids across membranes to generate lipid asymmetry, a property vital to many cellular processes. Mutations in several P4-ATPases have been linked to severe neurodegenerative and metabolic disorders. Most P4-ATPases associate with
Yoshiki Moriizumi et al.
Scientific reports, 8(1), 11757-11757 (2018-08-08)
We developed a novel hybrid cell reactor system via functional fusion of single Escherichia coli protoplast cells, that are deficient in cell wall and expose plasma membrane, with arrayed lipid bilayer chambers on a device in order to incorporate the
Tatjana Michel et al.
Molecular therapy. Nucleic acids, 8, 459-468 (2017-09-18)
Synthetically modified mRNA is a unique bioactive agent, ideal for use in therapeutic applications, such as cancer vaccination or treatment of single-gene disorders. In order to facilitate mRNA transfections for future therapeutic applications, there is a need for the delivery
Juliette Fedry et al.
PLoS biology, 16(8), e2006357-e2006357 (2018-08-14)
HAPLESS2 (HAP2) is a broadly conserved, gamete-expressed transmembrane protein that was shown recently to be structurally homologous to viral class II fusion proteins, which initiate fusion with host cells via insertion of fusion loops into the host membrane. However, the
Guangsheng Du et al.
Journal of controlled release : official journal of the Controlled Release Society, 266, 109-118 (2017-09-26)
In this study, we investigated the potential of intradermal delivery of nanoparticulate vaccines to modulate the immune response of protein antigen using hollow microneedles. Four types of nanoparticles covering a broad range of physiochemical parameters, namely poly (lactic-co-glycolic) (PLGA) nanoparticles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service