Skip to Content
Merck
All Photos(1)

Key Documents

P0430

Sigma-Aldrich

Periodic acid

suitable for electrophoresis, ≥99%

Synonym(s):

Iodic acid, Orthoperiodic acid, Paraperiodic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H5IO6
CAS Number:
Molecular Weight:
227.94
EC Number:
MDL number:
UNSPSC Code:
41105319
PubChem Substance ID:
NACRES:
NB.22

Assay

≥99%

form

powder or crystals

reaction suitability

reagent type: oxidant

technique(s)

electrophoresis: suitable

mp

122 °C (lit.)

solubility

water: 100 mg/mL, clear, colorless

SMILES string

OI(O)(O)(O)(O)=O

InChI

1S/H5IO6/c2-1(3,4,5,6)7/h(H5,2,3,4,5,6,7)

InChI key

TWLXDPFBEPBAQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Periodic acid is commonly used as a component for glycoprotein stains. Selective staining of glycoproteins on SDS-PAGE and membranes is carried out using a modification of the Periodic Acid-Schiff (PAS) method. Staining of sugar moieties of glycoproteins yields magenta bands with a colorless background. The Periodic Acid/Schiff Reagent stains vicinal diol groups found mainly on peripheral sugars and sialic acids and is used as a general glycoprotein stain. Periodic acid is also used to cleave reversible cross-linkers (e.g. DATD, DHEBA) in PAGE gels.

Analysis Note

Tested for use in staining of glycoproteins.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 1 - Skin Corr. 1B - STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christina Ernst et al.
Nature communications, 10(1), 1251-1251 (2019-03-21)
Male gametes are generated through a specialised differentiation pathway involving a series of developmental transitions that are poorly characterised at the molecular level. Here, we use droplet-based single-cell RNA-Sequencing to profile spermatogenesis in adult animals and at multiple stages during
Identification of glycoproteins on nitrocellulose membranes and gels.
D J Thornton et al.
Methods in molecular biology (Clifton, N.J.), 32, 119-128 (1994-01-01)
Glycoprotein staining following electrophoresis on acrylamide gels.
R M Zacharius et al.
Analytical biochemistry, 30(1), 148-152 (1969-07-01)
Courtney M Anderson et al.
Skeletal muscle, 5, 7-7 (2015-03-20)
Skeletal muscle is the most abundant tissue in the body and is a major source of total energy expenditure in mammals. Skeletal muscle consists of fast and slow fiber types, which differ in their energy usage, contractile speed, and force
Murad Alturkustani et al.
Journal of neuropathology and experimental neurology, 74(3), 233-240 (2015-02-11)
The pathologic features of adult-onset leukoencephalopathy/leukodystrophy with axonal spheroids (ALAS) are variable, and this has led to different hypotheses as to whether primarily demyelination or axonopathy may underlie this disorder. Typical ALAS pathology is rarely accompanied by focal multiple sclerosis

Articles

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service