Skip to Content
Merck
All Photos(3)

Key Documents

M8892

Sigma-Aldrich

Maltitol

≥98% (HPLC)

Synonym(s):

4-O-α-Glucopyranosyl-D-sorbitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24O11
CAS Number:
Molecular Weight:
344.31
Beilstein:
89983
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (HPLC)

form

powder

sweetness

0.9 × sucrose

color

white

mp

149-152 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

OC[C@H](O)[C@@H](O)[C@H](O[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H](O)CO

InChI

1S/C12H24O11/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12/h4-21H,1-3H2/t4-,5+,6+,7+,8+,9-,10+,11+,12+/m0/s1

InChI key

VQHSOMBJVWLPSR-WUJBLJFYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Maltitol is a low-caloric artificial sweetener, consisting of a sugar alcohol (polyol),

Application

Maltitol has been used in a study to assess the dissolution rate enhancement of piroxicam by ordered mixing. It has also been used in a study to investigate the comparative effects of the maltitol chewing gums on reducing plaque.

Biochem/physiol Actions

Sugar substitute with a weak genotoxic effect.

Other Notes

To gain a comprehensive understanding of our extensive range of Sugar alcohols for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative effects of the maltitol chewing gums on reducing plaque
Li, X., et al.
Hua xi Kou Qiang yi Xue za Zhi = Huaxi Kouqiang Yixue Zazhi = West China Journal of Stomatology, 57, 502-504 (2010)
Vikas Anand Saharan et al.
Pakistan journal of pharmaceutical sciences, 25(3), 521-533 (2012-06-21)
Micronized piroxicam was mixed with lactose, mannitol, sorbitol, maltitol and sodium chloride to produce ordered mixture in a glass vial by manual hand shaking method. The effect of excipients, surfactant, superdisintegrant, drug concentration and carrier particle size on dissolution rate
Semir Canimoglu et al.
Toxicology and industrial health, 29(10), 935-943 (2012-05-16)
In the present study, the genotoxic and cytotoxic effects of the low-caloric artificial sweetener maltitol, which is a sugar alcohol (polyol), were investigated in the bone marrow cells of rats using the chromosome aberration (CA) test. In addition, the teratogenicity
Vasso Psimouli et al.
Journal of the science of food and agriculture, 92(1), 99-105 (2011-08-13)
The aim of this research was to investigate whether certain polyols (mannitol, maltitol, sorbitol, lactitol), fructose, oligofructose and polydextrose can replace sugar (by an equal amount of each substitute) in cake formulations. The rheological behaviour of the cake batter and
Sergey Vyazovkin et al.
Pharmaceutical research, 23(9), 2158-2164 (2006-09-05)
To demonstrate the utility of differential scanning calorimetry (DSC) for determining activation energy landscape in amorphous pharmaceutical systems throughout the sub-Tg and Tg regions. DSC was employed to determine the effective activation energies (E) of the relaxation in sub-Tg and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service