Skip to Content
Merck
All Photos(1)

Key Documents

M7292

Sigma-Aldrich

M16 Medium

With sodium bicarbonate and lactic acid, without penicillin and streptomycin, liquid, sterile-filtered

Synonym(s):

Cell Culture Medium, Cell Culture Medium M16, M16 Medium Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | embryo: suitable

impurities

endotoxin, tested

components

NaHCO3: 2.101 g/L
phenol red: 0.0106 g/L
sodium pyruvate: 0.0363 g/L
glucose: 1.0 g/L (Dextro)

shipped in

ambient

storage temp.

2-8°C

Application

M2 and M16 Medium are common media for in vitro culture of preimplantation stage embryos. It is a modified Krebs-Ringer bicarbonate solution, which is very similar to Whitten′s Medium. M16 contains pyruvate and lactate as energy sources since preimplantation embryos do not utilize glucose efficiently.

Quality

Presence of particulates in solution will not affect product′s performance.

Reconstitution

Supplement with 0.06 g/L potassium penicillin-G and 0.05 g/L streptomycin sulfate.

Analysis Note

This product is tested for its ability to support the development of one-cell mouse embryos to expanded blastocysts. Minimum requirement is 80% development to blastocyst.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian-Qian Sha et al.
Nature communications, 9(1), 3477-3477 (2018-08-30)
Trimethylation of histone H3 on lysine-4 (H3K4me3) is associated with gene-regulatory elements, but its transcription-independent function in cell division is unclear. CxxC-finger protein-1 (CFP1) is a major mediator of H3K4 trimethylation in mouse oocytes. Here we report that oocyte-specific knockout
Neil Cw Mackenzie et al.
Journal of RNAi and gene silencing : an international journal of RNA and gene targeting research, 6(2), 416-421 (2011-02-26)
Transgenic technologies can provide important animal models for studying drug-metabolizing enzymes. Our overall aim was to generate versatile cell and animal systems that exhibited varying levels of cytochrome P450 oxidoreductase (POR) activity, more accurately modelling the human population for pharmacological
Kengo Horie et al.
Biochemical and biophysical research communications, 463(4), 907-911 (2015-06-14)
Prairie vole (Microtus ochrogaster) is a highly social animal and is a commonly used animal model for neuropsychopharmacological and psychiatric studies. To date, only a few reports on the development of transgenic prairie voles which was primarily due to the
Xinyan Cao et al.
International journal of molecular sciences, 20(9) (2019-05-01)
Embryo implantation in the mink follows the pattern of many carnivores, in that preimplantation embryo diapause occurs in every gestation. Details of the gene expression and regulatory networks that terminate embryo diapause remain poorly understood. Illumina RNA-Seq was used to
Hong-Bin Liu et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 6(15), 1900295-1900295 (2019-08-14)
A number of genes involved in zygotic genome activation (ZGA) have been identified, but the RNA-binding maternal factors that are directly related to ZGA in mice remain unclear. The present study shows that maternal deletion of Igf  2bp2 (also commonly known

Articles

Mouse embryo media and embryo validated reagents for transgenic mouse embryo culture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service