Skip to Content
Merck
All Photos(2)

Key Documents

M0925

Sigma-Aldrich

3α,6α-Mannopentaose

≥85%

Synonym(s):

α-Man-(1→3)(α-Man-[1→6])-α-Man-(1→6)(α-Man-[1→3])-Man

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H52O26
CAS Number:
Molecular Weight:
828.72
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥85%

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H](OC[C@H]2O[C@H](OC[C@@H](O)[C@@H](O)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]3O)[C@H](O)C=O)[C@@H](O)[C@@H](O[C@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]4O)[C@@H]2O)[C@@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C30H52O26/c31-1-7(35)25(55-29-22(46)19(43)15(39)10(3-33)52-29)13(37)8(36)5-49-28-24(48)26(56-30-23(47)20(44)16(40)11(4-34)53-30)17(41)12(54-28)6-50-27-21(45)18(42)14(38)9(2-32)51-27/h1,7-30,32-48H,2-6H2/t7-,8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18+,19+,20+,21+,22+,23+,24+,25-,26+,27+,28+,29-,30-/m1/s1

InChI key

YPQGRSSAXDUGEC-LHCUGFEVSA-N

General description

Core structure of some triantennary N-linked oligosaccharides

Application

3α,6α-Mannopentaose has been used:
  • in the modification of six valent osmium complexes with N,N,N′,N′-tetramethylethylenediamine (Os(VI)tem) for voltammetric analysis
  • to study the structure of Burkholderia oklahomensis EO147 agglutinin (BOA)
  • as a commercial isomeric oligosaccharide in matrix-assisted laser desorption/ionization (MALDI) LTQ orbitrap fragmentation studies

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fragmentation of neutral oligosaccharides using the MALDI LTQ Orbitrap
Rohmer M, et al.
International Journal of Mass Spectrometry, 305(2-3), 199-208 (2011)
Matthew J Whitley et al.
The FEBS journal, 280(9), 2056-2067 (2013-03-14)
Burkholderia oklahomensis EO147 agglutinin (BOA) is a 29 kDa member of the Oscillatoria agardhii agglutinin (OAA) family of lectins. Members of the OAA family recognize high-mannose glycans, and, by binding to the HIV envelope glycoprotein 120 (gp120), block the virus
Mojmír Trefulka et al.
Bioelectrochemistry (Amsterdam, Netherlands), 133, 107494-107494 (2020-03-03)
Compounds containing vicinal diol (glycol) groups, including saccharides, could be modified with sixvalent osmium complexes with nitrogenous ligands, particularly with N,N,N',N'-tetramethylethylenediamine (Os(VI)tem). The modification products are electrochemically active. Here we show that aminosaccharides can also be modified by Os(VI)tem. We

Articles

N-linked glycosylation, modification, and degradation

N-linked glycosylation, modification, and degradation

N-linked glycosylation, modification, and degradation

N-linked glycosylation, modification, and degradation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service