Skip to Content
Merck
All Photos(1)

Key Documents

M0268

Sigma-Aldrich

Minimum Essential Medium Eagle

With Earle′s salts and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture

Synonym(s):

EMEM, MEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

components

HEPES: no
phenol red: yes
Earle’s salts (5% CO2): yes
NaHCO3: no
L-glutamine: yes
sodium pyruvate: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Minimum Essential Medium Eagle (MEM) is one of the most widely used synthetic cell culture media. MEM has been used for culturing several cells grown in monolayers. It is commonly used to grow attached cell lines, such as fibroblasts, in the presence of fetal bovine sera (FBS), calf, or horse sera.

Application

Minimum Essential Medium Eagle has been used:
  • in the isolation and culture of primary hepatocytes
  • to culture the human endometrial carcinoma cells Ishikawa (ATTC) and L-Wnt-3a cells (ATCC) that secrete Wnt family member 3a (Wnt-3a) ligand
  • to culture the AN3CA (endometrial cancer cell line) cells, and HEC1-B human (endometrial adenocarcinoma cell line) cells

Features and Benefits

  • Minimum Essential Medium Eagle (MEM) contains higher concentrations of amino acids to mimic the protein composition of cultured mammalian cells.
  • The formulation has been altered by optional elimination of calcium to enable the growth of cells in suspension culture.

Quantity

Formulated to contain 9.5 grams of powder per liter of medium.

Reconstitution

Supplement with 2.2 g/L sodium bicarbonate.

Other Notes

Essentially the same as the original formula, except it is provided as a powder.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G F Shiguemoto et al.
Brazilian journal of biology = Revista brasleira de biologia, 81(2), 301-308 (2020-07-23)
Wild fish domestication can be considered a strategic approach to endangered species conservation, supporting studies and reducing economic and environmental costs. Three of the most important strategies in the domestication processes of fish are the adaptation of wild fish to
Jifu Mao et al.
ACS nano, 11(10), 10409-10416 (2017-09-08)
The poor mechanical properties and processability of pristine heterocyclic conductive polymers represent the most notable scientific and technological challenges that have greatly limited the application of these polymers. We report a soft and mechanically processable free-standing pristine polypyrrole (PPy) membrane
T L Freeman et al.
Biochemical pharmacology, 68(2), 223-230 (2004-06-15)
Activation of hepatic stellate cells (HSCs) involves the induction of ECM protein synthesis and rapid cell proliferation. Thus, agents that interfere with either process could potentially mitigate the development of liver disease by reducing the synthesis of proteins associated with
Enriqueta Garcia-Gutierrez et al.
PloS one, 11(3), e0150687-e0150687 (2016-03-02)
The metabolism of S. Typhimurium within infected host cells plays a fundamental role in virulence since it enables intracellular proliferation and dissemination and affects the innate immune response. An essential requirement for the intracellular replication of S. Typhimurium is the
Xiaolong Sun et al.
Acta histochemica, 122(8), 151649-151649 (2020-11-10)
As one of the most frequent diabetic complications, diabetic foot ulcer (DFU) can cause limb ischemia or even amputation. Paeoniflorin (PF) has been reported to possess many kinds of biological functions, such as antioxidant and anti-inflammatory effects. However, the role

Articles

A large selection of MEM formulations. Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.

A large selection of MEM formulations. Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.

A large selection of MEM formulations. Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.

A large selection of MEM formulations. Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.

Protocols

Powdered media and salt mixtures are extremely hygroscopic and should be protected from atmospheric moisture.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service