Skip to Content
Merck
All Photos(4)

Key Documents

L9037

Sigma-Aldrich

L-Lysine monohydrate

98.5-101.5% dry basis, suitable for cell culture, BioReagent, non-animal source

Synonym(s):

L-Lysine hydrate (1:1), Lysine monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2O2 · H2O
CAS Number:
Molecular Weight:
164.20
Beilstein:
4349963
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

L-Lysine monohydrate, BioReagent, suitable for cell culture, from non-animal source

biological source

non-animal source

product line

BioReagent

Assay

98.5-101.5% dry basis

form

crystalline powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white

mp

215 °C

solubility

H2O: soluble

SMILES string

[H]O[H].NCCCC[C@H](N)C(O)=O

InChI

1S/C6H14N2O2.H2O/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H2/t5-;/m0./s1

InChI key

HZRUTVAFDWTKGD-JEDNCBNOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Lysine is an amino acid with a basic side chain and is amphipathic in nature. It has a protonated alkyl amino group and is fully protonated at physiological pH. L-lysine mainly participates in electrostatic interactions in proteins.

Application

L-Lysine monohydrate has been used in:
  • SILAC (stable isotope labeling by amino acids in cell culture) method
  • the complete synthetic (C) medium for positive selection screening of yeast cells
  • the growth medium for cells

Biochem/physiol Actions

L-Lysine is an essential proteinogenic α amino acid used in a wide range of applications including as a non-animal sourced supplement in cell culture media, a substrate of enzymes such as L-lysine oxidase (EC 1.4.3.14), a component of poly-lysine polymers, and a substrate for oxidation and glycation mechanism studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shao-En Ong et al.
Nature protocols, 1(6), 2650-2660 (2007-04-05)
Stable isotope labeling by amino acids in cell culture (SILAC) is a simple, robust, yet powerful approach in mass spectrometry (MS)-based quantitative proteomics. SILAC labels cellular proteomes through normal metabolic processes, incorporating non-radioactive, stable isotope-containing amino acids in newly synthesized
Daniel Kalb et al.
Chembiochem : a European journal of chemical biology, 16(10), 1426-1430 (2015-05-13)
L-α-Aminoadipic acid reductases catalyze the ATP- and NADPH-dependent reduction of L-α-aminoadipic acid to the corresponding 6-semialdehyde during fungal L-lysine biosynthesis. These reductases resemble peptide synthetases with regard to their multidomain composition but feature a unique domain of elusive function--now referred
W L McKeehan et al.
Proceedings of the National Academy of Sciences of the United States of America, 73(6), 2023-2027 (1976-06-01)
The trace element selenium is essential for clonal growth of diploid fibroblasts from human fetal lung (WI-38) in media containing small amounts of serum protein. Maximum growth stimulation is obtained when 30 nM neutralized selenious acid is added to a
Garrett RH and Grisham CM
Biochemistry (2016)
Benjamin Newcomb et al.
Methods in molecular biology (Clifton, N.J.), 548, 145-160 (2009-06-13)
A multitude of enzymes that modify histones and remodel nucleosomes are required for the formation, maintenance, and propagation of the transcriptionally repressed chromatin state in eukaryotes. Robust phenotypic screens in yeast S. cerevisiae have proved instrumental in identifying these activities

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service