Skip to Content
Merck
All Photos(2)

Key Documents

L5918

Sigma-Aldrich

Lomustine

≥98%

Synonym(s):

1-(2-Chloroethyl)-3-cyclohexyl-1-nitrosourea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16ClN3O2
CAS Number:
Molecular Weight:
233.70
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98%

storage temp.

−20°C

SMILES string

ClCCN(N=O)C(=O)NC1CCCCC1

InChI

1S/C9H16ClN3O2/c10-6-7-13(12-15)9(14)11-8-4-2-1-3-5-8/h8H,1-7H2,(H,11,14)

InChI key

GQYIWUVLTXOXAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Antineoplastic agent with cellular DNA effects. Lomustine induces p53 expression in A2870 cells.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tracy T Batchelor et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(26), 3212-3218 (2013-08-14)
A randomized, phase III, placebo-controlled, partially blinded clinical trial (REGAL [Recent in in Glioblastoma Alone and With Lomustine]) was conducted to determine the efficacy of cediranib, an oral pan-vascular endothelial growth factor (VEGF) receptor tyrosine kinase inhibitor, either as monotherapy
C F Saba et al.
Veterinary and comparative oncology, 10(4), 283-291 (2012-01-13)
Treatment of feline vaccine-associated sarcoma (VAS) is challenging, in part due to the high likelihood of tumour recurrence despite aggressive local therapy. Lomustine is potentially an attractive agent to add to the current treatment armamentarium. In this de-escalating phase I/II
Daisuke Hasegawa et al.
The Journal of veterinary medical science, 74(11), 1517-1521 (2012-07-13)
A 9 year-old, neutered, male French Bulldog showing cluster seizures was diagnosed with a glioma in the right piriform cortex by MRI. Hypofractionated radiation therapy (RT) was performed using a linear accelerator. Although the lesion had involuted significantly at 2
Carsten Friedrich et al.
European journal of cancer (Oxford, England : 1990), 49(4), 893-903 (2012-11-28)
Medulloblastoma in adulthood is rare. Knowledge is limited, and the efficacy and toxicity of chemotherapy--especially in nonmetastatic disease--is still elusive. Seventy adults aged ≥21 years (median age: 28.5 years) with nonmetastatic medulloblastoma were followed as observational patients within the prospective
Joann L Ater et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 30(21), 2641-2647 (2012-06-06)
PURPOSE Surgery is curative therapy for pediatric low-grade gliomas (LGGs) in areas of the brain amenable to complete resection. However, LGGs located in areas where complete resection is not possible can threaten both function and life. The purpose of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service