Skip to Content
Merck
All Photos(1)

Key Documents

L3885

Sigma-Aldrich

Succinyl-Concanavalin A

lyophilized powder

Synonym(s):

Succ-Con A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

form

lyophilized powder

Quality Level

composition

Protein, ~90% E1%/280

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Succinyl Con A is the active dimeric (divalent) form which does not re-aggregate to form tetramers above pH 5.6. Agglutination activity is greatly reduced by the succinylation procedure, but reactivity with mannose or glucose residues is retained.

Packaging

Packaged based on protein content.

Quality

Highly purified.

Physical form

Contains sodium phosphate

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service