Skip to Content
Merck
All Photos(1)

Key Documents

L0520

Sigma-Aldrich

Lactoferrin from human milk

≥85% (SDS-PAGE), lyophilized powder

Synonym(s):

Growth-inhibiting protein 12, Lactotransferrin, Talalactoferrin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

human milk

Assay

≥85% (SDS-PAGE)

form

lyophilized powder

mol wt

82.4 kDa

composition

Protein, ≥90% biuret

technique(s)

microbiological culture: suitable

solubility

phosphate buffer: 1 mg/mL, clear to slightly hazy (0.01 M phosphate buffer, 0.0027 M potassium chloride and 0.137 M sodium chloride, pH 7.4, at 25 °C)

UniProt accession no.

storage temp.

2-8°C

InChI

1S/C35H53N9O12/c1-16(2)27(43-33(53)23(14-26(47)48)41-25(46)15-38-29(49)18(5)39-31(51)21(36)13-24(37)45)34(54)40-19(6)30(50)42-22(12-20-10-8-7-9-11-20)32(52)44-28(17(3)4)35(55)56/h7-11,16-19,21-23,27-28H,12-15,36H2,1-6H3,(H2,37,45)(H,38,49)(H,39,51)(H,40,54)(H,41,46)(H,42,50)(H,43,53)(H,44,52)(H,47,48)(H,55,56)/t18-,19-,21-,22-,23-,27-,28-/m0/s1

InChI key

QCBUWCQOKPLTDZ-PKRULZLPSA-N

Gene Information

human ... LTF(4057)

Looking for similar products? Visit Product Comparison Guide

Application

Lactoferrin was used to grow Streptococcus mutans in an iron-limiting medium. It was used to test if lactoferrin impedes epithelial cell adhesion in vitro.

Biochem/physiol Actions

Lactoferrin is an iron binding protein. It is structurally similar to transferrin, the plasma iron transport protein; but lactoferrin has a much higher affinity for iron (250 fold). It is very abundant in colostrum and small amounts can also be found in tears, saliva, mucous secretions and in the secondary granules of neutrophils. It is made by mucosal epithelium and neutrophils and is released by these cells in response to inflammatory stimuli. Bacterial growth is inhibited by its ability to sequester iron and also permeabilize bacterial cell walls by binding to lipopolysaccharides through its N-terminus. Lactoferrin can inhibit viral infection by binding tightly to the viral envelope protein. This prevents cell-virus fusion by blocking the binding domain. Lactoferrin appears to activate host defense systems in part by stimulating the release of interleukin-8, a neutrophil activator. It may also be involved in antibody and interleukin synthesis, lymphocyte proliferation and complement activation.

Quality

Contains iron, see certificate of analysis for lot specific information.

Preparation Note

Chromatographically purified.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liisa Kautto et al.
Experimental eye research, 145, 278-288 (2016-02-07)
The human eye is constantly bathed by tears, which protect the ocular surface via a variety of mechanisms. The O-linked glycans of tear mucins have long been considered to play a role in binding to pathogens and facilitating their removal
Shelton W Wright et al.
PLoS neglected tropical diseases, 14(8), e0008495-e0008495 (2020-08-09)
Melioidosis is an often-severe tropical infection caused by Burkholderia pseudomallei (Bp) with high associated morbidity and mortality. Burkholderia thailandensis (Bt) is a closely related surrogate that does not require BSL-3 conditions for study. Lactoferrin is an iron-binding glycoprotein that can
Michał Zimecki et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 61(11), 1247-1255 (2012-07-20)
The aim of this study was to assess the utility of lactoferrin (LF), a natural immunomodulator, to restrain allergen-induced pleurisy in mice. BALB/c female mice, 8- to 10-week old, weighing 24 g on average, were used. Mice were immunized intraperitoneally
Jessica R Sheldon et al.
PLoS pathogens, 16(10), e1008995-e1008995 (2020-10-20)
Acinetobacter baumannii is an emerging pathogen that poses a global health threat due to a lack of therapeutic options for treating drug-resistant strains. In addition to acquiring resistance to last-resort antibiotics, the success of A. baumannii is partially due to
M T Pöllänen et al.
Journal of periodontal research, 33(1), 8-16 (1998-04-03)
In the process of host defence against microbial challenge, neutrophils release granule contents with the potential side effect of damaging structural tissues. In the junctional epithelium such damage may contribute to the degeneration and renewal of the epithelial cells attached

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service