Skip to Content
Merck
All Photos(4)

Key Documents

I7253

Sigma-Aldrich

Isomaltose

~98% (TLC)

Synonym(s):

6-O-α-D-Glucopyranosyl-D-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
93355
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

~98% (TLC)

Quality Level

form

lyophilized powder

technique(s)

thin layer chromatography (TLC): suitable

color

white to yellow cast

solubility

water: 49.00-51.00 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H](OC[C@H]2OC(O)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-3-5(14)8(17)10(19)12(23-3)21-2-4-6(15)7(16)9(18)11(20)22-4/h3-20H,1-2H2/t3-,4-,5-,6-,7+,8+,9-,10-,11?,12+/m1/s1

InChI key

DLRVVLDZNNYCBX-RTPHMHGBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Comprehensive glucobioses review: A critical review discusses the properties, production, and diverse applications of functional glucobioses including isomaltose, emphasizing its role in glycemic index research and its potential as a sugar substitute in diabetic care (Zhang et al., 2023).

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Judith G P van Can et al.
The British journal of nutrition, 108(7), 1210-1217 (2011-12-17)
The impact of slowly digestible sugars in reducing the risk of developing obesity and related metabolic disorders remains unclear. We hypothesised that such carbohydrates (CHO), resulting in a lower glycaemic and insulinaemic response, may lead to greater postprandial fat oxidation
Kentaro Nakamura et al.
Clinical nutrition (Edinburgh, Scotland), 30(4), 513-516 (2011-02-02)
Gut ischemia-reperfusion (I/R) is considered an important mechanism underlying multiple organ failure after severe surgical insults. We previously demonstrated an enteral diet enriched with whey-hydrolyzed peptide, fermented milk, and isomaltulose to have anti-inflammatory effects in a concanavalin A-induced hepatitis model.
Jeff R Broadbent et al.
BMC genomics, 13, 533-533 (2012-10-06)
The broad ecological distribution of L. casei makes it an insightful subject for research on genome evolution and lifestyle adaptation. To explore evolutionary mechanisms that determine genomic diversity of L. casei, we performed comparative analysis of 17 L. casei genomes
John J Dubé et al.
Medicine and science in sports and exercise, 44(5), 793-799 (2011-11-05)
Exercise improves insulin resistance and is a first line for the prevention and treatment of type 2 diabetes. The extent, however, to which these responses are dose dependent is not known. The purpose of this study was to examine whether
Michael J Coffey et al.
The Cochrane database of systematic reviews, 1, CD012949-CD012949 (2020-01-22)
Cystic fibrosis (CF) is a multisystem disease and the importance of growth and nutrition has been well established, given its implications for lung function and overall survival. It has been established that intestinal dysbiosis (i.e. microbial imbalance) and inflammation is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service