Skip to Content
Merck
All Photos(1)

Key Documents

I5635

Sigma-Aldrich

Monoclonal Anti-Human IgG2 antibody produced in mouse

clone HP-6014, ascites fluid

Synonym(s):

Monoclonal Anti-Human IgG2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

secondary antibodies

clone

HP-6014, monoclonal

contains

15 mM sodium azide

technique(s)

indirect ELISA: 1:5,000

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-Human IgG2 (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Human IgG consists of four subclasses.

Specificity

The antibody is specific for human IgG2 and is non-reactive with other IgG subclasses. This clone has been singled out as the most widely applicable IgG2 specific monoclonal antibody by the IUIS/WHO study.

Application

Monoclonal Anti-Human IgG2 antibody has been used in in enzyme linked immunosorbent assay (ELISA) and in western blotting.
It may also be used:
  • in imprint immunofixation
  • in immunofluorometric assay
  • in hemagglutination
  • in hemagglutination inhibition
  • in particle counting immunoassay
  • in detection of cytoplasmic IgG

Biochem/physiol Actions

Anti-polysaccharide responses are mainly of the IgG2 sub-class while protein antigens give rise to IgG1 and IgG3 antibodies. Lipopolysaccharides stimulate an IgG2 response in peripheral blood leukocytes (PBL′s).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Negative immunodiffusion test results obtained with sera of paracoccidioidomycosis patients may be related to low-avidity immunoglobulin G2 antibodies directed against carbohydrate epitopes
Neves AR, et al.
Clinical and Diagnostic Laboratory Immunology, 10(5), 802-807 (2003)
Z Lazarova et al.
The Journal of investigative dermatology, 114(1), 178-184 (2000-01-05)
Patients with one form of cicatricial pemphigoid have IgG antibasement membrane autoantibodies against laminin 5 (alpha3beta3gamma2). Although passive transfer of rabbit anti-laminin 5 IgG to neonatal mice has been shown to induce subepidermal blisters that mimic those in patients, it
Identification of immune biomarkers related to disease progression and treatment efficacy in human visceral leishmaniasis
Portela A S B, et al.
Immunobiology, 223(3), 303-309 (2018)
A conserved Leishmania hypothetical protein evaluated for the serodiagnosis of canine and human visceral and tegumentary leishmaniasis, as well as a serological marker for the posttreatment patient follow-up
Ribeiro P A, et al.
Diagnostic Microbiology and Infectious Disease, 92(3), 196-203 (2018)
Takamasa Iwakura et al.
Internal medicine (Tokyo, Japan), 55(18), 2663-2668 (2016-09-16)
A 30-year-old woman with proteinuria first noted at 26 weeks of gestation was admitted to undergo further evaluation. A renal biopsy revealed membranous nephropathy (MN). There was no evidence of any secondary MN. Prednisolone was initiated 6 months after delivery.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service